Merck
  • Angiotensin II enhances endothelin-1-induced vasoconstriction through upregulating endothelin type A receptor.
Angiotensin II enhances endothelin-1-induced vasoconstriction through upregulating endothelin type A receptor.
Biochemical and biophysical research communications (2014-08-05)
Yan-Jie Lin, Ching-Fai Kwok, Chi-Chang Juan, Yung-Pei Hsu, Kuang-Chung Shih, Chin-Chang Chen, Low-Tone Ho
ZUSAMMENFASSUNG
Endothelin-1 (ET-1) is the most potent vasoconstrictor by binding to endothelin receptors (ETAR) in vascular smooth muscle cells (VSMCs). The complex of angiotensin II (Ang II) and Ang II type one receptor (AT1R) acts as a transient constrictor of VSMCs. The synergistic effect of ET-1 and Ang II on blood pressure has been observed in rats; however, the underlying mechanism remains unclear. We hypothesize that Ang II leads to enhancing ET-1-mediated vasoconstriction through the activation of endothelin receptor in VSMCs. The ET-1-induced vasoconstriction, ET-1 binding, and endothelin receptor expression were explored in the isolated endothelium-denuded aortae and A-10 VSMCs. Ang II pretreatment enhanced ET-1-induced vasoconstriction and ET-1 binding to the aorta. Ang II enhanced ETAR expression, but not ETBR, in aorta and increased ET-1 binding, mainly to ETAR in A-10 VSMCs. Moreover, Ang II-enhanced ETAR expression was blunted and ET-1 binding was reduced by AT1R antagonism or by inhibitors of PKC or ERK individually. In conclusion, Ang II enhances ET-1-induced vasoconstriction by upregulating ETAR expression and ET-1/ETAR binding, which may be because of the AngII/Ang II receptor pathways and the activation of PKC or ERK. These findings suggest the synergistic effect of Ang II and ET-1 on the pathogenic development of hypertension.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Urethan, ≥99%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Urethan, ≥99.0% (GC)
Sigma-Aldrich
SP600125, ≥98% (HPLC)
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Urethan, analytical standard