Merck
  • Circumvention of cisplatin resistance in ovarian cancer by combination of cyclosporin A and low-intensity ultrasound.
Circumvention of cisplatin resistance in ovarian cancer by combination of cyclosporin A and low-intensity ultrasound.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2015-02-11)
Tinghe Yu, Yan Yang, Jiao Zhang, Haining He, Xueyi Ren
ZUSAMMENFASSUNG
Cisplatin resistance is a challenge in the treatment of ovarian cancer. The aim of this study was to explore if ultrasound can overcome chemoresistance and enhance chemosensitization due to cyclosporin A. Ultrasound and/or cyclosporin A were employed to overcome cisplatin resistance in human ovarian cancer cell line COC1/DDP. Mechanisms were explored from the perspective of: DNA damage, intracellular platinum level, detoxification, and genes related to drug efflux and DNA repair. In vivo therapeutic efficacy was validated in a short-term model (subrenal cell-clot transplantation) in mice and the survival benefit was investigated in an orthotopic cancer model in mice using HO-8910PM cells. The findings were: (i) ultrasound enhanced the effect of cisplatin leading to a lower cell-survival rate (IC50 decreased from 3.19 to 0.35 μg/ml); (ii) ultrasound enhanced cisplatin via direct (increasing the intercellular level of active platinum) and indirect (decreasing the glutathione level, and expression of LRP and ERCC1 genes) mechanisms that intensified cisplatin-induced DNA damage, thus enhancing cell apoptosis and necrosis; (iii) cisplatin followed by ultrasound led to small tumor sizes in the short-term model without exacerbation of the systemic toxicity, and prolonged the survival times in the orthotopic model; and (iv) ultrasound synergized the sensitization due to cyclosporin A in vitro and in vivo. These data demonstrated that ultrasound combined with cyclosporin A overcame cisplatin resistance in ovarian cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
cis-Diamminplatin(II)-dichlorid, crystalline
Sigma-Aldrich
1-Chlor-2,4-dinitrobenzol, ≥99%
Sigma-Aldrich
Propidiumjodid -Lösung
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Sigma-Aldrich
1-Chlor-2,4-dinitrobenzol, 97%
Supelco
Glutathion, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutathion reduziert, BioXtra, ≥98.0%
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
Propidiumjodid, ≥94% (HPLC)
Supelco
Cyclosporin, Pharmaceutical Secondary Standard; Certified Reference Material
Cisplatin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
cis-Diaminplatin(II)-dichlorid, ≥99.9% trace metals basis
Sigma-Aldrich
Cyclosporin A, BioReagent, from Tolypocladium inflatum, Molecular Biology, ≥95%
Glutathion, European Pharmacopoeia (EP) Reference Standard
Supelco
Cyclosporin A, VETRANAL®, analytical standard
Sigma-Aldrich
trans-Platin(II)diammindichlorid
Cisplatin Unreinheit A, European Pharmacopoeia (EP) Reference Standard
USP
Transplatin, United States Pharmacopeia (USP) Reference Standard