Merck
  • Tanshinone IIA inhibits gastric carcinoma AGS cells through increasing p-p38, p-JNK and p53 but reducing p-ERK, CDC2 and cyclin B1 expression.
Tanshinone IIA inhibits gastric carcinoma AGS cells through increasing p-p38, p-JNK and p53 but reducing p-ERK, CDC2 and cyclin B1 expression.
Anticancer research (2014-12-17)
Chin-Cheng Su
ZUSAMMENFASSUNG
Tanshinone IIA (Tan-IIA) is extracted from Danshen (Salviae miltiorrhizae radix). It possesses antitumor activity against a variety of human cancer cells and its induction of apoptosis and inhibition of proliferation of gastric cancer cells are well-documented. However, the molecular mechanisms by which Tan-IIA inhibits gastric cancer have not been well-elucidated. In the present study, we evaluated the cytotoxicity of Tan-IIA against human gastric cancer AGS cells by the (3-(4, 5-dimethylthiazol-2-yl)-2, 5-diphenyltetrazolium bromide) MTT assay. The protein expression of tumor necrosis factor-alpha (TNF-α), FAS, p53, p21, cyclin A, cyclin B1, extracellular-related kinase (ERK), phospho extracellular-related kinase (p-ERK), p38, p-p38, Jun-amino-terminal kinase (JNK), phospho Jun-amino-terminal kinase (p-JNK) and β-actin in AGS cells were measured by western blotting. The cell-cycle distribution was analyzed by flow cytometry. The results showed that Tan-IIA inhibited AGS cells with time- and dose-dependent manners. AGS cells treated with Tan-IIA up-regulated the protein expression of TNFα, FAS, p-p38, p-JNK, p53, p21, caspase-3 and caspase-8 but reduced that of p-ERK, CDC2, cyclin A, and cyclin B1. The results also showed that Tan-IIA dose dependently induced G2/M phase arrest. These findings demonstrate that Tan-IIA can inhibit AGS human gastric cancer cells; one of the molecular mechanisms may be through increasing the protein expression of p-p38 and p-JNK, but decreasing that of p-ERK to induce the activation of p53, followed by increasing the protein expression of p21 to down-regulate CDC2 and cyclin B1 expression which then induces G2/M phase arrest. Another route may be through increasing the protein expression of TNF-α, FAS, caspase-8 and caspase-3 to induce apoptosis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Deoxycholsäure Natriumsalz, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Deoxycholsäure Natriumsalz, ≥97% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
Sigma-Aldrich
Kaliumphosphat, tribasisch, reagent grade, ≥97%
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
SAFC
Deoxycholsäure Natriumsalz