Merck
Hydrogen sulfide improves glucose metabolism and prevents hypertrophy in cardiomyocytes.
Nitric oxide : biology and chemistry (2014-12-20)
Min Liang, Sheng Jin, Dong-Dong Wu, Ming-Jie Wang, Yi-Chun Zhu
ZUSAMMENFASSUNG
Hydrogen sulfide (H2S) has been reported to inhibit myocardial hypertrophy in a cell model of cardiomyocyte hypertrophy. Our previous study also shows an H2S-induced increase in glucose metabolism in insulin-targeting cells. The present study aims to examine the hypothesis that H2S attenuates myocardial hypertrophy and promotes glucose utilization in cardiomyocytes. The cell model of cardiomyocyte hypertrophy was induced by application of phenylephrine and cardiomyocyte hypertrophy was examined using leucine incorporation assay. Protein levels were measured using Western blot analysis. The activity of related enzymes was measured with enzyme-linked immunosorbent assay (ELISA). NaHS (an H2S donor) treatment increased the activity of cultured cardiomyocytes and reduced hypertrophy in cultured cardiomyocytes at concentrations ranging from 25 to 200 µmol/L. NaHS treatment increased glucose uptake and the efficiency of glycolysis and the citric acid cycle. The key enzymes in these reactions, including lactate dehydrogenase and pyruvate kinase and succinate dehydrogenase, were activated by NaHS treatment (100 µmol/L). Some intermediates of glycolysis and the citric acid cycle, including lactic acid, cyclohexylammonium, oxaloacetic acid, succinate, L-dimalate, sodium citrate, cis-aconitic acid, ketoglutarate and DL-isocitric acid trisodium also showed anti-hypertrophic effects in cardiomyocytes. H2S improves glucose utilization and inhibits cardiomyocyte hypertrophy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Milchsäure, meets USP testing specifications
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Milchsäure, 88%, FCC
Sigma-Aldrich
Oxalessigsäure, ≥97% (HPLC)
Sigma-Aldrich
Milchsäure, natural, ≥85%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Milchsäure -Lösung, ACS reagent, ≥85%
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
cis-Aconitsäure, ≥98%
Supelco
Milchsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Milchsäure, ~90% (T)
Sigma-Aldrich
5-Brom-2′-deoxyuridin, BioUltra, ≥99%
Sigma-Aldrich
Oxalessigsäure, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97% (HPLC)
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%