Merck
Rapid dissolution of propofol emulsions under sink conditions.
International journal of pharmaceutics (2015-02-01)
Robert Damitz, Anuj Chauhan
ZUSAMMENFASSUNG
Pain accompanying intravenous injections of propofol is a major problem in anesthesia. Pain is ascribed to the interaction of propofol with the local vasculature and could be impacted by rapid dissolution of the emulsion formulation to release the drug. In this paper, we measure the dissolution of propofol emulsions including the commercial formulation Diprivan(®). We image the turbidity of blood protein sink solutions after emulsions are injected. The images are digitized, and the drug release times are estimated from the pixel intensity data for a range of starting emulsion droplet size. Drug release times are compared to a mechanistic model. After injection, pixel intensity or turbidity decreases due to reductions in emulsion droplet size. Drug release times can still be measured even if the emulsion does not completely dissolve such as with Diprivan(®). Both pure propofol emulsions and Diprivan(®) release drug very rapidly (under five seconds). Reducing emulsion droplet size significantly increases the drug release rate. Drug release times observed are slightly longer than the model prediction likely due to imperfect mixing. Drug release from emulsions occurs very rapidly after injection. This could be a contributing factor to pain on injection of propofol emulsions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Natriumoctanoat, ≥99% (capillary GC)
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
2,6-Diisopropylphenol, 97%
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Propofol, European Pharmacopoeia (EP) Reference Standard
Propofol für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard