Merck
  • Ostertagia ostertagi macrophage migration inhibitory factor is present in all developmental stages and may cross-regulate host functions through interaction with the host receptor.
Ostertagia ostertagi macrophage migration inhibitory factor is present in all developmental stages and may cross-regulate host functions through interaction with the host receptor.
International journal for parasitology (2014-03-04)
Guanggang Qu, Raymond Fetterer, Lin Leng, Xin Du, Dante Zarlenga, Zhiqiang Shen, Wenyu Han, Richard Bucala, Wenbin Tuo
ZUSAMMENFASSUNG
Macrophage migration inhibitory factor (MIF) of Ostertagia ostertagi, an abomasal parasite of cattle, was characterised in the present study. Phylogenetic analysis identified at least three O. ostertagi MIFs (Oos-MIFs), each encoded by a distinct transcript: Oos-MIF-1.1, Oos-MIF-1.2 and Oos-MIF-2. Oos-MIF-2 is only distantly related to Oos-MIF-1s, but has higher sequence similarity with the Caenorhabditis elegans MIF2. Oos-MIF-1.1 and Oos-MIF-1.2 are similar (93%) and thus collectively referred to as Oos-MIF-1 when characterised with immunoassays. Recombinant Oos-MIF-1.1 (rOos-MIF-1.1) is catalytically active as a tautomerase. A mutation (rOos-MIF-1.1P1G) or duplication of Pro1 residue (rOos-MIF-1.1P1+P) resulted in reduced oligomerisation and loss of tautomerase activity. The tautomerase activity of rOos-MIF-1.1 was only partially inhibited by ISO-1 but was abrogated by a rOos-MIF-1.1-specific antibody. Oos-MIF-1 was detected in all developmental stages of O. ostertagi, with higher levels in the adult stage; it was also detected in adult worm excretory/secretory product. Oos-MIF-1 was localised to the hypodermis/muscle, reproductive tract and intestine, but not to the cuticle. rOos-MIF-1.1, but not rOos-MIF-1.1P1G, was able to specifically bind to human CD74, a MIF cell surface receptor, with an affinity comparable with human MIF. Immunostaining indicated that macrophages were able to internalise rOos-MIF-1.1, further supporting receptor-mediated transportation. Herein we also show that rOos-MIF-1.1 inhibited migration of bovine macrophages and restored glucocorticoid-suppressed, lipopolysaccharide-induced TNF-α and IL-8 in human and/or bovine macrophages. Given its dual role in self-regulation and molecular mimicry, this secreted parasite protein warrants investigation as a vaccine candidate against O. ostertagi infections in cattle.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumperiodat, ACS reagent, ≥99.8%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Natrium-(Meta-)Periodat, ≥99.0%
Sigma-Aldrich
Trichloressigsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Deoxycholsäure Natriumsalz, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Deoxycholsäure Natriumsalz, ≥97% (titration)
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Trichloressigsäure -Lösung, 6.1 N
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyd -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Trichloressigsäure, BioUltra, ≥99.5% (T)
SAFC
Deoxycholsäure Natriumsalz
Sigma-Aldrich
Trichloressigsäure, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Trichloressigsäure, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloressigsäure, ≥99.0% (titration)
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyd -Lösung, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutarsäure-Dialdehyd -Lösung, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyd -Lösung, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Natrium-(Meta-)Periodat, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Trichloressigsäure, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%