Merck
  • Semi-interpenetrating network (sIPN) co-electrospun gelatin/insulin fiber formulation for transbuccal insulin delivery.
Semi-interpenetrating network (sIPN) co-electrospun gelatin/insulin fiber formulation for transbuccal insulin delivery.
Pharmaceutical research (2014-07-18)
Leyuan Xu, Natasha Sheybani, Shunlin Ren, Gary L Bowlin, W Andrew Yeudall, Hu Yang
ZUSAMMENFASSUNG
This work was aimed at developing a semi-interpenetrating network (sIPN) co-electrospun gelatin/insulin fiber scaffold (GIF) formulation for transbuccal insulin delivery. Gelatin was electrospun into fibers and converted into an sIPN following eosin Y-initiated polymerization of polyethylene glycol diacrylate (PEG-DA). The cytocompatibility, degradation rate and mechanical properties were examined in the resulting sIPNs with various ratios of PEG-DA to eosin Y to find a suitable formulation for transbuccal drug delivery. Insulin was co-electrospun with gelatin into fibers and converted into an sIPN-GIF using this suitable formulation. The in vitro release kinetics of insulin was evaluated using ELISA. The bioactivity of released insulin was analyzed in 3T3-L1 preadipocytes using Western blotting and Oil Red O staining. The transbuccal permeability of released insulin was determined using an in vitro porcine oral mucosa model. The sIPN-GF formulation of GF cross-linked by PEG-DA (1% w/v) with eosin Y (5% v/v) possessed no cytotoxic effect, a moderate degradation rate with degradation half-life of 49 min, and a significant enhancement in mechanical properties. This formulation was used to fabricate sIPN-GIF. Insulin release was extended up to 4 h by sIPN-GIF. The released insulin successfully triggered intracellular AKT phosphorylation and induced adipocyte differentiation in 3T3-L1 preadipocytes. The transbuccal permeability of released insulin was determined on the order of 10(-7) cm/s. Insulin can be fabricated into an sIPN-GIF formulation following co-electrospinning and cross-linking without losing bioactivity. It proved the potential of this new formulation for transbuccal insulin delivery.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
1,1,1,3,3,3-Hexafluor-2-propanol, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
3-Isobutyl-1-methylxanthin, ≥99% (HPLC), powder
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
1-Vinyl-2-pyrrolidon, contains sodium hydroxide as inhibitor, ≥99%
Sigma-Aldrich
Triethanolamin, ≥99.0% (GC)
Sigma-Aldrich
3-Isobutyl-1-methylxanthin, BioUltra, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Sigma-Aldrich
Triethanolamin, reagent grade, 98%
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol, standard for GC
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Eosin Y -Lösung, 5 wt. % in H2O
Sigma-Aldrich
Eosin Y, Dye content ~99 %
Sigma-Aldrich
Eosin Y Dinatriumsalz, certified by the BSC
Supelco
Dexamethason, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
1,1,1,3,3,3-Hexafluor-2-propanol, derivatization grade (GC derivatization), LiChropur, ≥99.8%
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Eosin Y Dinatriumsalz, ≥85 % by HPLC
Sigma-Aldrich
Ethanol
Sigma-Aldrich
Eosin Y, 75% (HPLC)
Supelco
Ethanol -Lösung, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Triethanolamin, BioUltra, ≥99.5% (GC)