Merck
  • Ameliorative effects of 5-hydroxymethyl-2-furfural (5-HMF) from Schisandra chinensis on alcoholic liver oxidative injury in mice.
Ameliorative effects of 5-hydroxymethyl-2-furfural (5-HMF) from Schisandra chinensis on alcoholic liver oxidative injury in mice.
International journal of molecular sciences (2015-01-27)
Wei Li, Xin-Nan Qu, Ye Han, Si-Wen Zheng, Jia Wang, Ying-Ping Wang
ZUSAMMENFASSUNG
The aim of this paper is to evaluate the protective effect of 5-hydroxymethyl-2-furfural (5-HMF) on acute alcohol-induced liver oxidative injury in mice. 5-HMF, a maillard reaction product, was isolated from the fruits of Schisandra chinensis for animal experiments. Experimental ICR mice were pretreated with different doses of 5-HMF (7.5, 15, and 30 mg/kg) for seven days by gavage feeding. Biochemical markers and enzymatic antioxidants from serum and liver tissue were examined. Our results showed that the activities of ALT (alanine aminotransferase), AST (aspartate transaminase), TC (total cholesterol), TG (triglyceride), L-DLC (low density lipoprotein) in serum and the levels of MDA (malondialdehyde) in liver tissue, decreased significantly (p < 0.05) in the 5-HMF-treated group compared with the alcohol group. On the contrary, enzymatic antioxidants CAT (catalase), GSH-Px (glutathione peroxidase), and GSH SOD (superoxide dismutase) were markedly elevated in liver tissue treated with 5-HMF (p < 0.05). Furthermore, the hepatic levels of pro-inflammatory response marker tumor necrosis factor-alpha (TNF-α) and interleukin-1β (IL-1β) were significantly suppressed (p < 0.05). Histopathological examination revealed that 5-HMF (30 mg/kg) pretreatment noticeably prevented alcohol-induced hepatocyte apoptosis and fatty degeneration. It is suggested that the hepatoprotective effects exhibited by 5-HMF on alcohol-induced liver oxidative injury may be due to its potent antioxidant properties.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Supelco
Methanol, analytical standard
Sigma-Aldrich
4,4′-Diaminodiphenylmethan, ≥97.0% (GC)
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Supelco
Glutathion, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutathion reduziert, BioXtra, ≥98.0%
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
DL-Alanin, ≥99% (HPLC)
Sigma-Aldrich
DL-Alanin, ≥99%, FCC, FG
Glutathion, European Pharmacopoeia (EP) Reference Standard
Supelco
4,4′-Diaminodiphenylmethan, analytical standard
Alanin, European Pharmacopoeia (EP) Reference Standard