Merck
  • Comparative evaluation of eight software programs for alignment of gas chromatography-mass spectrometry chromatograms in metabolomics experiments.
Comparative evaluation of eight software programs for alignment of gas chromatography-mass spectrometry chromatograms in metabolomics experiments.
Journal of chromatography. A (2014-12-02)
Weihuan Niu, Elisa Knight, Qingyou Xia, Brian D McGarvey
ZUSAMMENFASSUNG
Since retention times of compounds in GC-MS chromatograms always vary slightly from chromatogram to chromatogram, it is necessary to align chromatograms before comparing them in metabolomics experiments. Several software programs have been developed to automate this process. Here we report a comparative evaluation of the performance of eight programs using prepared samples of mixtures of chemicals, and an extract of tomato vines spiked with three concentrations of a mixture of alkanes. The programs included in the comparison were SpectConnect, MetaboliteDetector 2.01a, MetAlign 041012, MZmine 2.0, TagFinder 04, XCMS Online 1.21.01, MeltDB and GAVIN. Samples were analyzed by GC-MS, chromatograms were aligned using the selected programs, and the resulting data matrices were preprocessed and submitted to principal components analysis. In the first trial, SpectConnect, MetAlign and MetaboliteDetector correctly identified ≥90% of the true positives. In the second trial, MetAlign and MetaboliteDetector correctly identified 87% and 81% of the true positives, respectively. In addition, in both trials >90% of the peaks identified by MetAlign and MetaboliteDetector were true positives.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichlormethan, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Hexan, Laboratory Reagent, ≥95%
Supelco
Hexan, analytical standard
Supelco
Dichlormethan, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dichlormethan, analytical standard
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichlormethan, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Dichlormethan, Selectophore, ≥99.5%