Merck
  • Sirt3 protects cortical neurons against oxidative stress via regulating mitochondrial Ca2+ and mitochondrial biogenesis.
Sirt3 protects cortical neurons against oxidative stress via regulating mitochondrial Ca2+ and mitochondrial biogenesis.
International journal of molecular sciences (2014-09-10)
Shu-Hui Dai, Tao Chen, Yu-Hai Wang, Jie Zhu, Peng Luo, Wei Rao, Yue-Fan Yang, Zhou Fei, Xiao-Fan Jiang
ZUSAMMENFASSUNG
Oxidative stress is a well-established event in the pathology of several neurobiological diseases. Sirt3 is a nicotinamide adenine nucleotide (NAD+)-dependent protein deacetylase that regulates mitochondrial function and metabolism in response to caloric restriction and stress. This study aims to investigate the role of Sirt3 in H2O2 induced oxidative neuronal injury in primary cultured rat cortical neurons. We found that H2O2 treatment significantly increased the expression of Sirt3 in a time-dependent manner at both mRNA and protein levels. Knockdown of Sirt3 with a specific small interfering RNA (siRNA) exacerbated H2O2-induced neuronal injury, whereas overexpression of Sirt3 by lentivirus transfection inhibited H2O2-induced neuronal damage reduced the generation of reactive oxygen species (ROS), and increased the activities of endogenous antioxidant enzymes. In addition, the intra-mitochondrial Ca2+ overload, but not cytosolic Ca2+ increase after H2O2 treatment, was strongly attenuated after Sirt3 overexpression. Overexpression of Sirt3 also increased the content of mitochondrial DNA (mtDNA) and the expression of mitochondrial biogenesis related transcription factors. All these results suggest that Sirt3 acts as a prosurvival factor playing an essential role to protect cortical neurons under H2O2 induced oxidative stress, possibly through regulating mitochondrial Ca2+ homeostasis and mitochondrial biogenesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Kaliumphosphat, tribasisch, reagent grade, ≥97%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Mangan, powder, ≥99.9% trace metals basis
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Mangan, powder, −325 mesh, ≥99% trace metals basis
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Mangan, chips, thickness <2.0 mm, 99%
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Mangan, foil, 25x25mm, thickness 2mm, hot-pressed, 99.95%
Mangan, foil, 50x50mm, thickness 2mm, hot-pressed, 99.95%
Mangan, foil, not light tested, 25x25mm, thickness 0.025mm, permanent polyester support, 98.7%
Mangan, rod, 10mm, diameter 4.0mm, cast, 99.5%
Mangan, rod, 50mm, diameter 4.0mm, cast, 99.5%