Merck
Stabilization and targeting of INO80 to replication forks by BAP1 during normal DNA synthesis.
Nature communications (2014-10-07)
Han-Sae Lee, Shin-Ai Lee, Shin-Kyoung Hur, Jeong-Wook Seo, Jongbum Kwon
ZUSAMMENFASSUNG
The INO80 chromatin-remodelling complex has been implicated in DNA replication during stress in yeast. However, its role in normal DNA replication and its underlying mechanisms remain unclear. Here, we show that INO80 binds to replication forks and promotes fork progression in human cells under unperturbed, normal conditions. We find that Ino80, which encodes the catalytic ATPase of INO80, is essential for mouse embryonic DNA replication and development. Ino80 is recruited to replication forks through interaction with ubiquitinated H2A--aided by BRCA1-associated protein-1 (BAP1), a tumour suppressor and nuclear de-ubiquitinating enzyme that also functions to stabilize Ino80. Importantly, Ino80 is downregulated in BAP1-defective cancer cells due to the lack of an Ino80 stabilization mechanism via BAP1. Our results establish a role for INO80 in normal DNA replication and uncover a mechanism by which this remodeler is targeted to replication forks, suggesting a molecular basis for the tumour-suppressing function of BAP1.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Kalium, chunks (in mineral oil), 98% trace metals basis
Sigma-Aldrich
β-tri-Calcium-phosphat, puriss. p.a., ≥98% β-phase basis (sintered Powder)
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Kaliumhydrid, 30 wt % dispersion in mineral oil
Sigma-Aldrich
β-tri-Calcium-phosphat, puriss. p.a., ≥98% β-phase basis (unsintered powder)
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Kaliumhydrid, in paraffin
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
αβ-Tri-Calciumphosphat, Reagent for transient & stable DNA transfections