Merck
  • Apoptotic and neurotoxic actions of 4-para-nonylphenol are accompanied by activation of retinoid X receptor and impairment of classical estrogen receptor signaling.
Apoptotic and neurotoxic actions of 4-para-nonylphenol are accompanied by activation of retinoid X receptor and impairment of classical estrogen receptor signaling.
The Journal of steroid biochemistry and molecular biology (2014-08-06)
E Litwa, J Rzemieniec, A Wnuk, W Lason, W Krzeptowski, M Kajta
ZUSAMMENFASSUNG
4-para-Nonylphenol (NP) is a non-ionic surfactant that has widespread and uncontrolled distribution in the environment. Little is known, however, about its actions on neuronal cells during critical developmental periods. This study aimed to investigate the mechanisms underlying the apoptotic and toxic actions of NP on mouse embryonic neuronal cells and the possible interactions of NP with estrogen receptor (ER)- and retinoid X receptor (RXR)-mediated intracellular signaling. Treatment of mouse hippocampal neuronal cell cultures with NP (5 and 10μM) induced apoptotic and neurotoxic effects. The 2 and 7 day-old mouse hippocampal cultures were vulnerable to 5 and 10μM NP, whereas 12 day-old cultures responded only to the highest concentration of NP, thus suggesting an age-dependent action of the chemical on neuronal cells. The use of specific inhibitors did not support the involvement of calpains in NP-induced apoptosis, but indicated caspase-8- and caspase-9-dependent effects of NP. Specific ER antagonists MPP and PHTPP potentiated the NP-induced loss of mitochondrial membrane potential and increase in lactate dehydrogenase (LDH) release whereas, ER agonists PPT and DPN inhibited these effects. RXR antagonist HX531 diminished the NP-evoked loss of mitochondrial membrane potential, the activity of caspase-3 and LDH release. In addition, exposure to NP inhibited ERα- and ERβ-specific immunofluorescence but stimulated RXR-specific immunolabeling in mouse hippocampal cells. In conclusion, our study demonstrated that the apoptotic and toxic actions of NP on neuronal cells in early development is accompanied by an impairment of ER- and stimulation of RXR-mediated signaling pathways. Taking into account NP-induced alterations in mRNA expression levels of particular types of RXRs, we suggest that NP affected mainly RXRα and RXRβ, but not RXRγ signaling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
DL-Dithiothreitol, Molecular Biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
DL-Dithiothreitol, BioUltra, Molecular Biology, ≥99.5% (RT)
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis