Merck
Three-dimensional analyses of a perforated cylindrical drug delivery device.
International journal of pharmaceutics (2015-02-01)
Laurent Simon, Juan Ospina
ZUSAMMENFASSUNG
A closed-formed solution of a perforated drug-delivery model was developed. Laplace transforms were applied to the governing equation, which included diffusion through the tubular device and mass transfer across a rectangular cut. A first-order estimate for the fraction of drug released, in terms of the Laplace variable, was derived after employing suitable boundary and initial conditions. The effective time constant for the process was calculated. The residue theorem and the Zakian method were proposed as two reliable approaches to recover the solution in the time domain. Simulations show that the drug was released faster at higher Sherwood numbers. Ninety-eight percent (98%) of the loading dose was delivered after a period corresponding to four time constants. This analytical platform can aid in the design of implants for long-term delivery applications.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Stearinsäure, Grade I, ≥98.5% (capillary GC)
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Stearinsäure, reagent grade, 95%
Sigma-Aldrich
Stearinsäure, ≥95%, FCC, FG
USP
Stearinsäure, United States Pharmacopeia (USP) Reference Standard
Supelco
Stearinsäure, analytical standard
Supelco
Stearinsäure, Pharmaceutical Secondary Standard; Certified Reference Material
Stearinsäure, European Pharmacopoeia (EP) Reference Standard
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Supelco
Stearinsäure, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland