Merck
  • Changes in the expression of miRNAs at the pericentral and periportal regions of the rat liver in response to hepatocellular injury: comparison with the changes in the expression of plasma miRNAs.
Changes in the expression of miRNAs at the pericentral and periportal regions of the rat liver in response to hepatocellular injury: comparison with the changes in the expression of plasma miRNAs.
Toxicology (2014-05-28)
Yu Yamaura, Miki Nakajima, Naoyuki Tatsumi, Shingo Takagi, Tatsuki Fukami, Koichi Tsuneyama, Tsuyoshi Yokoi
ZUSAMMENFASSUNG
MicroRNAs (miRNAs) in body fluids have received attention as potential biomarkers of organ damage because miRNAs that are highly or specifically expressed in a given organ are likely released into body fluids as a result of damage to that organ. We previously determined that the plasma miRNA profile in rats was dramatically changed due to acetaminophen (APAP)-induced pericentral necrosis and methapyrilene (MP)-induced periportal necrosis in the liver. The purpose of this study was to examine whether the expression of hepatic miRNAs is differentially modulated at different zones due to injury and to examine the relationship of the hepatic miRNA profile with the changes in the plasma miRNA expression profile. Through the laser microdissection of the periportal and periportal regions of the liver and TaqMan microRNA Array analysis, we found that 49 miRNAs are differentially expressed between the pericentral and periportal regions of control rats. In both APAP- and MP-treated rats, the miRNAs that presented decreased expression dominated in both the injured and non-injured areas compared with the miRNAs that exhibited increased expression. The changes in miRNA expression in each region of the liver were compared with those observed in the plasma. Of the 301 plasma miRNAs with expression that was changed as a result of APAP administration, only 21% were changed in the injured area of the liver. Of the 263 plasma miRNAs with expression that was changed due to MP administration, only 24% were changed in the injured area of the liver. Thus, the miRNA expression profiles in the plasma do not merely reflect the release of miRNAs from the damaged cells in the liver. This report provides the first demonstration of zonal miRNA expression in the liver and of the relationship of the miRNA expression profile in a tissue with the plasma miRNA profile.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
4-Acetamidophenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder
USP
Acetaminophen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
4-Acetamidophenol, BioXtra, ≥99.0%
Sigma-Aldrich
Hämatoxylin, certified by the BSC
Sigma-Aldrich
4-Acetamidophenol, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
4-Acetamidophenol, analytical standard
Paracetamol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®