Merck
  • Myricetin and naringenin inhibit human squamous cell carcinoma proliferation and migration in vitro.
Myricetin and naringenin inhibit human squamous cell carcinoma proliferation and migration in vitro.
Nutrition and cancer (2014-09-27)
Daniele Maggioni, Gabriella Nicolini, Roberta Rigolio, Luisa Biffi, Lorenzo Pignataro, Renato Gaini, Werner Garavello
ZUSAMMENFASSUNG
In this study the potential anticancer effect of 2 flavonoids, myiricetin (MYR) and naringenin (NAR) has been evaluated on an oral squamous cell carcinoma (OSCC) cell line, SCC-25, and HaCaT cells. Both the flavonoids inhibited SCC-25 cell growth, although NAR selectively affected cancer cells without impairing HaCaT cell growth. The cell proliferation inhibition by MYR and NAR was not related to apoptosis induction, but on cell cycle impairment, because a G0/G1 and a G2/M blockage was highlighted following 24 h of treatment in SCC-25 and HaCaT cells, respectively. Western blot analysis showed that MYR induced a decrease of Cyclin D1 in SCC-25 and of Cyclin B1 in HaCaT cells, while NAR negatively modulated Cyclin D1 expression in SCC-25 cells. Wound-healing and cell invasion assays demonstrated that both the flavonoids were able to reduce motility on both SCC-25 and HaCaT cells. In conclusion the results of the present study show the anticancer potential of NAR and MYR on OSCC because they exert cytostatic effect by the impairment of cell cycle progression. Moreover both the flavonoids inhibit cell migration, thus highlighting their potential effect as antimetastatic agents. Therefore, MYR and NAR appear as promising candidate as oral cancer chemopreventive agents.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
Hydrocortison, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, Molecular Biology, ≥97.0%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Hydrocortison, γ-irradiated, powder, BioXtra, suitable for cell culture