Merck
Effect of surface functionality of silica nanoparticles on cellular uptake and cytotoxicity.
Molecular pharmaceutics (2014-08-29)
Siddharth Jambhrunkar, Zhi Qu, Amirali Popat, Jie Yang, Owen Noonan, Luiz Acauan, Yusilawati Ahmad Nor, Chengzhong Yu, Surajit Karmakar
ZUSAMMENFASSUNG
Mesoporous silica nanoparticles (MCM-41) with different surface chemistry were used as carrier system to study its influence on drug delivery and anticancer activity of curcumin (CUR). CUR was encapsulated in pristine MCM-41 (hydrophilic and negatively charged), amino functionalized MCM-41 (MCM-41-NH2 which is hydrophilic and positively charged), and methyl functionalized MCM-41 (MCM-41-CH3 which is hydrophobic and negatively charged) and evaluated for in vitro release and cell cytotoxicity in human squamous cell carcinoma cell line (SCC25). Various techniques were employed to evaluate the performance of these materials on cellular uptake and anticancer activity in the SCC25 cell line. Both positively and negatively charged surfaces demonstrated enhanced drug release and anticancer activity compared to pure CUR. Positively charged nanoparticles showed higher cell uptake compared to negatively charged nanoparticles owing to its electrostatic interaction with cells. However, hydrophobic surface modified nanoparticles (MCM-41-CH3) showed no improvement in drug release and anticancer activity due to its poor wetting effect. Cell cycle analysis and cell apoptosis studies revealed different pathway mechanisms followed by the positively and negatively charged nanoparticles but exhibiting similar anticancer activity in SCC25 cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Tetraethylorthosilikat, reagent grade, 98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilan, 99%
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, ≥98%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Propidiumjodid, ≥94.0% (HPLC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Hexadecyltrimethylammoniumbromid, Molecular Biology, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Chlorotrimethylsilan, ≥98.0% (GC)
Sigma-Aldrich
Tetraethylorthosilikat, ≥99.0% (GC)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Curcumin, from Curcuma longa (Turmeric), powder
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Chlorotrimethylsilan, purified by redistillation, ≥99%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)