Merck
  • CCL18 promotes epithelial-mesenchymal transition, invasion and migration of pancreatic cancer cells in pancreatic ductal adenocarcinoma.
CCL18 promotes epithelial-mesenchymal transition, invasion and migration of pancreatic cancer cells in pancreatic ductal adenocarcinoma.
International journal of oncology (2014-12-17)
Fanbin Meng, Wan Li, Changling Li, Zhigang Gao, Kejian Guo, Shaowei Song
ZUSAMMENFASSUNG
CCL18 is a chemokine that is primarily expressed in monocytes, macrophages and immature dendritic cells and plays a crucial role in immune and inflammation responses. Recently, CCL18 was found to play pivotal roles in the development of several kinds of cancers, but its expression status and role during the tumorigenesis of pancreatic cancer remain unknown. In this study, we performed immunohistochemistry and enzyme-linked immunosorbent assay (ELISA) to evaluate the expression of CCL18 in human pancreatic ductal adenocarcinoma (PDAC) tissues and preoperative serum, respectively. The results showed that both cancer epithelial cells and mesenchymal macrophages in PDAC tissues positively expressed CCL18. Serum CCL18 levels were significantly higher in patients with PDAC in comparison to healthy controls. The expression of CCL18 in both cancer epithelial cells and mesenchymal cells was correlated with lymph node metastasis, histopathological grading and overall survival in 62 PDAC patients. In vitro assays showed that the gene and protein expression of CCL18 from U937 and THP-1 cell- derived macrophages were significantly higher than that from unstimulated U937 cells and THP-1 cells. In contrast, pancreatic cancer cell lines showed little to no CCL18 expression even after IL4 stimulation. Intriguingly, pancreatic cancer cell lines expressed the potential CCL18 receptors PITPNM3, CCR6 and GPR3. Furthermore, treatment with recombinant human CCL18 promoted the migration and invasion of pancreatic cancer cells, but had no effect on cell proliferation. Consistent with these results, CCL18 induced the expression of the epithelial-mesenchymal transition (EMT) related gene SNAIL1. Our findings suggest that the serum level of CCL18 is a potential biomarker for the diagnosis and prognosis of PDAC, and that the combined functions of CCL18 in mesenchymal and cancer cells might accelerate the progression of PDAC by promoting the epithelial-mesenchymal transition, invasion and migration of pancreatic cancer cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Wasser, suitable for HPLC
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Wasser, Deionized
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Wasser, suitable for ion chromatography
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Wasser, for TOC analysis
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Wasser, BioPerformance Certified
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Wasser-Dichtestandard, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Wasser, ACS reagent
Supelco
Wasser, for ultratrace analysis, ACS reagent
Sigma-Aldrich
Wasser, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Wasser, PCR Reagent