Merck
Alpha-lipoic acid activates eNOS through activation of PI3-kinase/Akt signaling pathway.
Vascular pharmacology (2014-12-03)
Zhekang Ying, Xiaoyun Xie, Minjie Chen, Kevin Yi, Sanjay Rajagopalan
ZUSAMMENFASSUNG
Lipoic acid (LA) exerts therapeutic effects on cardiovascular diseases. However, the mechanisms underlying these therapeutic effects remain elusive. Endothelial nitric oxide synthase (eNOS) plays a critical role in cardiovascular homeostasis. LA was shown to potently activate PI3-kinase/Akt pathway, and the latter is critical in the regulation of eNOS activity. In the present study, we test the hypothesis that LA improves endothelial function through PI3-kinase/Akt-mediated eNOS activation. Western blot analysis showed that LA time- and dose-dependently induced phosphorylation of Akt and eNOS in human umbilical vein endothelial cells (HUVECs). Both PI3-kinase and Akt inhibitors abolished LA-induced eNOS phosphorylation, indicating that LA induces eNOS phosphorylation through the PI3-kinase/Akt pathway. This increase in eNOS phosphorylation was paralleled by an increase in NO release by HUVECs, supporting its relevance in eNOS activity regulation. Myograph analysis revealed that LA relaxed phenylephrine-induced contraction. Endothelium removal and NOS inhibition by L-NAME abolished this vasodilator action of LA, and Akt but not AMPK inhibition significantly reduced the vasodilator action of LA, indicating that it is mediated by PI3-kinase/Akt pathway-dependent activation of eNOS. Consistent with in vitro results, intraperitoneal injection with LA significantly increased plasma nitrite and nitrate levels in C57Bl/6j mice. LA activates eNOS through a PI3-kinase/Akt signaling pathway-dependent mechanism, offering a potential molecular basis for the therapeutic effects of LA on cardiovascular diseases.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumfluorid, ACS reagent, ≥99%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
Deoxycholsäure Natriumsalz, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Deoxycholsäure Natriumsalz, ≥97% (titration)
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, Molecular Biology, ≥97.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Sigma-Aldrich
Natriumfluorid, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, ≥97.0%
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
SAFC
Deoxycholsäure Natriumsalz
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Natriumfluorid, 99.99% trace metals basis
Sigma-Aldrich
DL-α-Liponsäure, synthetic, ≥99% (titration), powder
Supelco
Natriumfluorid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenglykol-bis(2-aminoethylether)-N,N,N′,N′-Tetraessigsäure, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumfluorid, anhydrous, powder, 99.99% trace metals basis