Merck
  • Assessment of the relative performance of a confined impinging jets mixer and a multi-inlet vortex mixer for curcumin nanoparticle production.
Assessment of the relative performance of a confined impinging jets mixer and a multi-inlet vortex mixer for curcumin nanoparticle production.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2014-07-16)
Shing Fung Chow, Changquan Calvin Sun, Albert Hee Lum Chow
ZUSAMMENFASSUNG
The relative performance of two specially designed mixers for nanoparticle production, namely, two-stream confined impinging jets with dilution mixer (CIJ-D-M) and four-stream multi-inlet vortex mixer (MIVM), was evaluated using the model compound, curcumin (CUR), under defined conditions of varying mixing rate and organic solvent. In the absence of turbulent fluctuations, higher mixing rate tended to generate finer particles. Among the three water-miscible organic solvents tested, acetone afforded the smallest particle size and the narrowest particle size distribution. Both mixers were capable of reproducibly fabricating CUR nanoparticles with particle size below 100 nm and high encapsulation efficiency (>99.9%). Specifically, CIJ-D-M yielded nanoparticles with smaller size and polydispersity index while the particles obtained by the MIVM displayed better short-term stability. In addition, CIJ-D-M tended to produce a mixture of irregular nanoaggregates and primary nanoparticles while roughly spherical nanoparticles were generated with the MIVM. The observed particle size and morphological differences could be attributed to the differences in the configuration of the mixing chamber and the related mixing order.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamid, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamid, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamid, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamid, biotech. grade, ≥99.9%
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Supelco
Dimethylformamid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Supelco
Tetrahydrofuran, analytical standard
Supelco
N,N-Dimethylformamid, analytical standard
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%