Merck
  • Comorbidity and high viral load linked to clinical presentation of respiratory human bocavirus infection.
Comorbidity and high viral load linked to clinical presentation of respiratory human bocavirus infection.
Archives of virology (2014-10-02)
Lucía María Ghietto, Diego Majul, Patricia Ferreyra Soaje, Elsa Baumeister, Martín Avaro, Constanza Insfrán, Liliana Mosca, Alicia Cámara, Laura Beatriz Moreno, Maria Pilar Adamo
ZUSAMMENFASSUNG
Human bocavirus (HBoV) is a new parvovirus associated with acute respiratory tract infection (ARTI). In order to evaluate HBoV significance as an agent of acute respiratory disease, we screened 1,135 respiratory samples from children and adults with and without symptoms during two complete calendar years. HBoV1 prevalence in patients with ARTI was 6.33 % in 2011 and 11.64 % in 2012, including neonatal and adult patients. HBoV1 was also detected in 3.77 % of asymptomatic individuals. The co-detection rate was 78.1 %. Among children, 87 % were clinically diagnosed with lower respiratory infection (no significant differences between patients with and without coinfection), and 31 % exhibited comorbidities. Pediatric patients with comorbidities were significantly older than patients without comorbidities. Patients with ARTI had either high or low viral load, while controls had only low viral load, but there were no clinical differences between patients with high or low viral load. In conclusion, we present evidence of the pathogenic potential of HBoV1 in young children with ARTI. Since patients with HBoV1-single infection are not significantly different from those with coinfection with respect to clinical features, the virus can be as pathogenic by itself as other respiratory agents are. Furthermore, an association between high HBoV1 load and disease could not be demonstrated in this study, but all asymptomatic individuals had low viral loads. Also, children with comorbidities are susceptible to HBoV1 infection at older ages than previously healthy children. Thus, the clinical presentation of infection may occur depending on both viral load and the particular interaction between the HBoV1 and the host.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethidiumbromid -Lösung, BioReagent, Molecular Biology, 10 mg/mL in H2O
Sigma-Aldrich
Ethidiumbromid -Lösung, BioReagent, Molecular Biology, 500 μg/mL in H2O
Sigma-Aldrich
Ethidiumbromid, BioReagent, Molecular Biology, powder
Sigma-Aldrich
Ethidiumbromid, ~95% (HPLC)
Sigma-Aldrich
Ethidiumbromid -Lösung, suitable for fluorescence, ~1% in H2O