Merck
  • Rosiglitazone attenuates atherosclerosis and increases high-density lipoprotein function in atherosclerotic rabbits.
Rosiglitazone attenuates atherosclerosis and increases high-density lipoprotein function in atherosclerotic rabbits.
International journal of molecular medicine (2015-01-22)
Chen Li, Yan Tu, Ting-Rong Liu, Zhi-Gang Guo, Di Xie, Jian-Kai Zhong, Yong-Zhen Fan, Wen-Yan Lai
ZUSAMMENFASSUNG
Rosiglitazone has been found to have anti-atherogenic effects and to increase serum high-density lipoprotein (HDL) cholesterol (HDL-C) levels. However, in vivo studies investigating the regulation of adenosine triphosphate-binding cassette transporter A1 (ABCA1) and scavenger receptor class B type I (SR-BI) by rosiglitazone are limited. Moreover, the effects of rosiglitazone on the function and levels of HDL are unclear. In the present study, we investigated the effects of rosiglitazone on HDL function and its mechanisms of action in atherosclerotic rabbits. Our results revealed that rosiglitazone induced a significant increase in serum HDL-C levels, paraoxonase 1 (PON1) activity, [(3)H]cholesterol efflux rates, and the expression of ABCA1 and SR-BI in hepatocytes and peritoneal macrophages. The expression of ABCA1 was also increased in aortic lesions. Rosiglitazone markedly reduced serum myeloperoxidase (MPO) activity, aortic intima-media thickness (IMT) and the percentage of plaque area in the aorta. It can thus be concluded that in atherosclerotic rabbits, rosigitazone increases the levels of HDL-C and hinders atherosclerosis. Thus, it improves HDL quality and function, as well as the HDL-induced cholesterol efflux, exerting anti-inflammatory and antioxidant effects.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyd -Lösung, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Formaldehyd -Lösung, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Phenylacetat, 99%
Sigma-Aldrich
Formaldehyd -Lösung, tested according to Ph. Eur.
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Formaldehyd-12C -Lösung, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.