Merck
  • Serum coenzyme Q10 and risk of disabling dementia: the Circulatory Risk in Communities Study (CIRCS).
Serum coenzyme Q10 and risk of disabling dementia: the Circulatory Risk in Communities Study (CIRCS).
Atherosclerosis (2014-12-03)
Kazumasa Yamagishi, Ai Ikeda, Yuri Moriyama, Choy-Lye Chei, Hiroyuki Noda, Mitsumasa Umesawa, Renzhe Cui, Masanori Nagao, Akihiko Kitamura, Yorihiro Yamamoto, Takashi Asada, Hiroyasu Iso
ZUSAMMENFASSUNG
To examine whether coenzyme Q10, a potent antioxidant, is associated with risk of dementia, which has not yet been elucidated. We performed a case-control study nested in a community-based cohort of approximately 6000 Japanese aged 40-69 years at baseline (1984-1994). Serum coenzyme Q10 was measured in 65 incident cases of disabling dementia with dementia-related behavioral disturbance or cognitive impairment incident between 1999 and 2004, and in 130 age-, sex- and baseline year-matched controls. Serum coenzyme Q10 was inversely associated with dementia: the multivariate odds ratios (95% confidence intervals) were 0.68 (0.26-1.78), 0.92 (0.33-2.56), and 0.23 (0.06-0.86) for individuals with the second, third, and highest quartiles of coenzyme Q10, respectively, as compared with the lowest quartile (P for trend = 0.05). A similar association was found for the coenzyme Q10/total cholesterol ratio: the respective ORs were 0.67 (0.25-1.78), 0.73 (0.28-1.92), and 0.21 (0.05-0.90) (P for trend = 0.04). Serum coenzyme Q10 levels were inversely associated with risk of disabling dementia.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.