Merck
  • Efficient purification of CD4+ lymphocytes from peripheral blood progenitor cell products using affinity bead acoustophoresis.
Efficient purification of CD4+ lymphocytes from peripheral blood progenitor cell products using affinity bead acoustophoresis.
Cytometry. Part A : the journal of the International Society for Analytical Cytology (2014-07-24)
Andreas Lenshof, Arshad Jamal, Josefina Dykes, Anke Urbansky, Ingbritt Astrand-Grundström, Thomas Laurell, Stefan Scheding
ZUSAMMENFASSUNG
Processing of peripheral blood progenitor cells (PBPC) for clinical transplantation or research applications aims to effectively isolate or deplete specific cell populations, utilizing primarily magnetic or fluorescence activated sorting methods. Here, we investigated the performance of microfluidic acoustophoresis for the separation of lymphocyte subsets from PBPC, and present a novel method for affinity-bead-mediated acoustic separation of cells which can otherwise not be acoustically discriminated. As the acoustic force on a particle depends on particle size, density and compressibility, targeting of cells by affinity specific beads will generate cell-bead complexes that exhibit distinct acoustic properties relative to nontargeted cells and are, thus, possible to isolate. To demonstrate this, PBPC samples (n = 22) were obtained from patients and healthy donors. Following density gradient centrifugation, cells were labeled with anti-CD4-coated magnetic beads (Dynal) and isolated by acoustophoresis and, for comparison, standard magnetic cell sorting technique in parallel. Targeted CD4+ lymphocytes were acoustically isolated with a mean (±SD) purity of 87 ± 12%, compared with 96 ± 3% for control magnetic sorting. Viability of sorted cells was 95 ± 4% (acoustic) and 97 ± 3% (magnetic), respectively. The mean acoustic separation efficiency of CD4+ lymphocytes to the target fraction was 65 ± 22%, compared with a mean CD4+ lymphocyte recovery of 56 ± 15% for magnetic sorting. Functional testing of targeted CD4+ lymphocytes demonstrated unimpaired mitogen-mediated proliferation capacity and cytokine production. Hematopoietic progenitor cell assays revealed a preserved colony forming ability of nontarget cells post sorting. We conclude that the acoustophoresis platform can be utilized to efficiently isolate bead-labeled CD4+ lymphocytes from PBPC samples in a continuous flow format, with preserved functional capacity of both target and nontarget cells. These results open up for simultaneous affinity-bead-mediated separation of multiple cell populations, something which is not possible with current standard magnetic cell separation technology. © 2014 International Society for Advancement of Cytometry.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumazid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Natriumazid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Natriumazid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Natriumazid, BioXtra
Sigma-Aldrich
5(6)-Carboxyfluoreszeindiacetat-N-succinimidylester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
5-Carboxy-Fluoresceindiacetat N-Succinimidylester, suitable for fluorescence, ≥95.0% (HPLC)
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar