Merck
  • High affinity anti-Internalin B VHH antibody fragments isolated from naturally and artificially immunized repertoires.
High affinity anti-Internalin B VHH antibody fragments isolated from naturally and artificially immunized repertoires.
Journal of immunological methods (2014-12-03)
Robert W Gene, Jyothi Kumaran, Cristina Aroche, Henk van Faassen, J Christopher Hall, C Roger MacKenzie, Mehdi Arbabi-Ghahroudi
ZUSAMMENFASSUNG
The need for rapid and easy technologies for the detection of food-borne and environmental pathogens is essential for safeguarding the health of populations. Furthermore, distribution of tainted food and water can have consequences which can affect whole economies. Antibodies and antibody fragments have been historically used in detection platforms due to their antigen specificity and robust physicochemical properties. In this study, we report the isolation and characterization of antibody fragments from the heavy chain antibody repertoire (VHH) of Camelidae which bind with specificity and high affinity to the Listeria monocytogenes invasin, Internalin B (InlB). To the best of our knowledge, this is the first report of anti-InlB VHHs from camelids. These anti-InlB VHHs were not cross-reactive to the structurally related Listeria invasin Internalin A (InlA) and are potential reagents to be used in the development of detection and medical technologies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Imidazol, ReagentPlus®, 99%
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Imidazol, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Imidazol, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Phosphorsäure, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Imidazol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Methanol, Absolute - Acetone free