Merck
  • Remarkably regioselective deacylation of cellulose esters using tetraalkylammonium salts of the strongly basic hydroxide ion.
Remarkably regioselective deacylation of cellulose esters using tetraalkylammonium salts of the strongly basic hydroxide ion.
Carbohydrate polymers (2014-07-20)
Xueyan Zheng, Richard D Gandour, Kevin J Edgar
ZUSAMMENFASSUNG
Tetraalkylammonium hydroxides have been found to mediate regioselective deacylation of cellulose esters. This deacylation surprisingly shows substantial selectivity for the removal of the acyl groups at O-2/3, affording cellulose-6-O-esters by a simple, efficient one-step process. The mechanism for this deacylation was investigated by studying the effect of tetraalkylammonium cation size upon ester deacylation selectivity. We hypothesize that coordination of the tetraalkylammonium cation by the ester oxygen atoms of the vicinal 2,3-acetate groups may drive the unexpected regioselectivity at the secondary alcohol esters. Broad scope with respect to ester type was demonstrated; regioselective O-2,3 deacylation was observed with cellulose acetate, propionate, butyrate, hexanoate and benzoate triesters. The scope of this deacylation of cellulose acetates has been investigated to understand how to carry it out most efficiently. Reaction with TBAOH in pyridine was the most effective process, providing the highest selectivity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Pyridin, ACS reagent, ≥99.0%
Sigma-Aldrich
Lithiumchlorid, ACS reagent, ≥99%
Sigma-Aldrich
N,N-Dimethylacetamid, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
1,4-Dioxan, ACS reagent, ≥99.0%, contains ≤25 ppm BHT as stabilizer
Sigma-Aldrich
N,N-Dimethylacetamid, ReagentPlus®, 99%
Sigma-Aldrich
Pyridin, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Pyridin, anhydrous, 99.8%
Sigma-Aldrich
4-(Dimethylamino)-pyridin, ReagentPlus®, ≥99%
Sigma-Aldrich
1,4-Dioxan, suitable for HPLC, ≥99.5%
Sigma-Aldrich
Tetrabutylammoniumhydroxid -Lösung, 40 wt. % in H2O
Sigma-Aldrich
N,N-Dimethylacetamid, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithiumchlorid, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, 99%
Sigma-Aldrich
N,N-Dimethylacetamid, anhydrous, 99.8%
Sigma-Aldrich
Lithiumchlorid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Tetrabutylammoniumhydroxid -Lösung, 1.0 M in methanol
Sigma-Aldrich
1,4-Dioxan, ACS reagent, ≥99.0%, contains <=25 ppm BHT as stabilizer
Sigma-Aldrich
Essigsäureanhydrid, ReagentPlus®, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Lithiumchlorid, ReagentPlus®, 99%
Sigma-Aldrich
Lithiumchlorid -Lösung, 8 M, Molecular Biology, ≥99%
Supelco
Tetrabutylammoniumhydroxid -Lösung, ~40% in water, suitable for ion chromatography
Sigma-Aldrich
Tetramethylammoniumhydroxid -Lösung, 25 wt. % in H2O