Merck
Amyrin attenuates scopolamine-induced cognitive impairment in mice.
Biological & pharmaceutical bulletin (2014-07-06)
Se Jin Park, Young Je Ahn, Sa Rang Oh, Younghwan Lee, Guyoung Kwon, Hyun Woo, Hyung Eun Lee, Dae Sik Jang, Ji Wook Jung, Jong Hoon Ryu
ZUSAMMENFASSUNG
Alzheimer's disease, a neurodegenerative disorder, is characterized by progressive cognitive impairment associated with the disruption of cholinergic neurotransmission. The aim of the present study was to evaluate the effect of α- or β-amyrin, a type of pentacyclic triterpene, on the cognitive impairment induced by scopolamine, a muscarinic acetylcholine receptor antagonist. To measure the abilities of various types of learning and memory, we conducted step-through passive avoidance task. Scopolamine induced deficits in learning and memory processes in mice, which were antagonized by a single administration of α-amyrin (2 or 4 mg/kg) or β-amyrin (4 mg/kg), respectively. Additionally, in vitro analysis revealed that acetylcholinesterase activity was inhibited by β-amyrin, but not by α-amyrin. Furthermore, Western blot analysis revealed that the expression levels of phosphorylated extracellular signal-regulated kinase 1/2 (pERK) and phosphorylated glycogen synthase kinase-3β (pGSK-3β) were significantly enhanced by a single administration of α- and β-amyrin in the hippocampus. Finally, the memory ameliorating effects of α- or β-amyrin on the scopolamine-induced cognitive impairments were significantly blocked by ERK inhibitor U0126. The present study suggests that α- and β-amyrin may ameliorate the cognitive impairment induced by hypocholinergic neurotransmission via the activation of ERK as well as GSK-3β signaling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Threonin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonin, reagent grade, ≥98% (HPLC)
SAFC
L-Threonin
Sigma-Aldrich
(−)-Scopolamin -hydrobromid Trihydrat, >95% (HPLC), powder
Sigma-Aldrich
Scopolamin -hydrobromid, meets USP testing specifications
Sigma-Aldrich
L-Threonin, BioXtra, ≥99.5% (NT)
Supelco
L-Threonin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Serin, ≥98% (TLC)
Tyrosin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
DL-Tyrosin, 99%
Sigma-Aldrich
DL-Serin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Serin, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Threonin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
L-Threonin, European Pharmacopoeia (EP) Reference Standard