Merck
  • Potential impact of metal oxide nanoparticles on the immune system: The role of integrins, L-selectin and the chemokine receptor CXCR4.
Potential impact of metal oxide nanoparticles on the immune system: The role of integrins, L-selectin and the chemokine receptor CXCR4.
Nanomedicine : nanotechnology, biology, and medicine (2014-03-22)
Tamara Lozano-Fernández, Lidia Ballester-Antxordoki, Nerea Pérez-Temprano, Elena Rojas, David Sanz, Maite Iglesias-Gaspar, Sergio Moya, África González-Fernández, Mercedes Rey
ZUSAMMENFASSUNG
The impact of metal oxide nanoparticles (NPs) on the immune system has been studied in vitro using human peripheral blood lymphocytes (PBLs). Metal oxide NPs (ZnO, CeO2, TiO2 and Al2O3) induced changes in the expression levels of adhesion molecules and the C-X-C chemokine receptor type 4 (CXCR4) in these cells. Proliferation studies were carried out with CFSE in response to PHA, finding an increase in T-cell proliferation upon cell exposure to TiO2 and Al2O3 NPs. For ZnO NPs, a decrease in the chemotactic response to SDF-1α was observed. No changes were found in basophil activation and leukocyte oxidative burst after phagocytosis. Despite the absence of cytotoxicity, metal oxide NPs are not inert; they alter the expression levels of adhesion molecules and chemokine receptors, key actors in the immune response, and affect important cell functions such as T-cell proliferative response to mitogens and chemotaxis. This study demonstrates the immune-modulating effects of four different metal nanoparticles in a human peripheral blood lymphocyte model system. These effects were clearly present even though these nanoparticles did not display cytotocity in ex vivo experiments.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
5(6)-Carboxyfluoreszeindiacetat-N-succinimidylester, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Supelco
Calcium Ionenlösung für ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
NGF-β human, from human, recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture
Sigma-Aldrich
5-Carboxy-Fluoresceindiacetat N-Succinimidylester, suitable for fluorescence, ≥95.0% (HPLC)