Merck
Investigating the anti-leishmanial effects of linear peptoids.
ChemMedChem (2014-12-17)
Gabriela A Eggimann, Hannah L Bolt, Paul W Denny, Steven L Cobb
ZUSAMMENFASSUNG
Peptoids, a class of peptide mimetics, have emerged as promising anti-infective agents against a range of bacterial infections. Herein we present the first study of the antiparasitic and specifically the anti-leishmanial properties of linear peptoids. Peptoids were identified as having promising activity against Leishmania mexicana axenic amastigotes, a causative agent of cutaneous leishmaniasis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
N,N-Dimethylformamid, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamid, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Sigma-Aldrich
N,N-Dimethylformamid, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamid, biotech. grade, ≥99.9%
Supelco
Trifluoressigsäure, analytical standard
Supelco
Dimethylformamid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Bromessigsäure, reagent grade, 97%
Supelco
N,N-Dimethylformamid, analytical standard
Sigma-Aldrich
Bromessigsäure, ReagentPlus®, ≥99.0%