Merck
Carnosic acid suppresses colon tumor formation in association with antiadipogenic activity.
Molecular nutrition & food research (2014-09-11)
Yun-Ji Kim, Jong-Sang Kim, Young Rok Seo, Jung Han Yoon Park, Myung-Sook Choi, Mi-Kyung Sung
ZUSAMMENFASSUNG
This study determined the efficacy of carnosic acid (CA) for suppressing colon carcinogenesis associated with excess adiposity. Cell growth regulation by CA was evaluated in HT-29 colon adenocarcinoma cells cocultured with 3T3-L1 adipocytes. To determine the in vivo efficacies, male A/J mice were divided into four groups and fed one of the following experimental diets for 11 wk: 15% fat, 45% fat, 45% fat + 0.01% CA, or 45% fat + 0.02% CA. Azoxymethane was administered at the beginning of experimental diet and two cycles of dextran sodium sulfate were supplied 1 wk after the azoxymethane injection. The proliferation of HT-29 cells cocultured with 3T3-L1 cells was significantly higher than proliferation of control cells (p < 0.05). CA treatment suppressed the growth of cocultured HT-29 cells through cell cycle arrest and enhanced apoptosis by inhibiting leptin receptor (Ob-R) signaling, including Akt and extracellular signal-regulated kinase (ERK) phosphorylation. CA supplementation in vivo decreased the number of colon tumors and reduced circulating concentrations of leptin, adiponectin, insulin, and insulin-like growth factor 1. Colonic expression of Ob-R, insulin receptor (IR), p-Akt, p-ERK, B-cell lymphoma extra large (Bcl-xL), and cyclinD1 protein was also suppressed in animals fed CA. CA appears to alleviate adipocity-related acceleration of colon tumor formation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Isopropylalkohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Isopropylalkohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Azoxymethan, 13.4 M, ≥98%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles