Merck
  • Spray drying formulation of albendazole microspheres by experimental design. In vitro-in vivo studies.
Spray drying formulation of albendazole microspheres by experimental design. In vitro-in vivo studies.
Drug development and industrial pharmacy (2013-11-19)
Agustina García, Darío Leonardi, Gisela N Piccirilli, María E Mamprin, Alejandro C Olivieri, María C Lamas
ZUSAMMENFASSUNG
Both an experimental design and optimization techniques were carried out for the development of chitosan-pectin-carboxymethylcellulose microspheres to improve the oral absorption of albendazole as a model drug. The effect of three different factors (chitosan, pectin and carboxy methyl cellulose concentrations) was studied on five responses: yield, morphology, dissolution rate at 30 and 60 min, and encapsulation efficiency of the microspheres. During the screening phase, the factors were evaluated in order to identify those which exert a significant effect. Simultaneous multiple response optimizations were then used to find out experimental conditions where the system shows the most adequate results. The optimal conditions were found to be: chitosan concentration, 1.00% w/v, pectin concentration 0.10% w/v and carboxymethylcellulose concentration 0.20% w/v. The bioavailability of the loaded drug in the optimized microspheres was evaluated in Wistar rats which showed an area under curve (AUC) almost 10 times higher than the pure drug.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fluorwasserstoffsäure, ACS reagent, 48%
Sigma-Aldrich
Ammoniumphosphat, einbasig, ACS reagent, ≥98%
Sigma-Aldrich
Fluorwasserstoffsäure, 48 wt. % in H2O, ≥99.99% trace metals basis
Sigma-Aldrich
Ammoniumphosphat, einbasig, ReagentPlus®, ≥98.5%
Sigma-Aldrich
Ammoniumphosphat, einbasig, ≥99.99% trace metals basis
Sigma-Aldrich
Ammoniumdihydrogenphosphat, 99.999% trace metals basis
Sigma-Aldrich
Ammoniumphosphat, einbasig, BioUltra, ≥99.5% (T)
Supelco
Ammoniumphosphat, einbasig, analytical standard, for nitrogen determination according to Kjeldahl method, ≥99.5%
Sigma-Aldrich
Ammoniumphosphat, einbasig, suitable for plant cell culture