Merck
Real-time monitoring of peptide grafting onto chitosan films using capillary electrophoresis.
Analytical and bioanalytical chemistry (2015-02-15)
Danielle L Taylor, Joel J Thevarajah, Diksha K Narayan, Patricia Murphy, Melissa M Mangala, Seakcheng Lim, Richard Wuhrer, Catherine Lefay, Michael D O'Connor, Marianne Gaborieau, Patrice Castignolles
ZUSAMMENFASSUNG
Chitosan, being antimicrobial and biocompatible, is attractive as a cell growth substrate. To improve cell attachment, arginine-glycine-aspartic acid-serine (RGDS) peptides were covalently grafted to chitosan films, through the widely used coupling agents 1-ethyl-3-(3-dimethylaminopropyl)carbodiimide (EDC-HCl) and N-hydroxysuccinimide (NHS), via the carboxylic acid function of the RGDS molecule. The grafting reaction was monitored, for the first time, in real time using free-solution capillary electrophoresis (CE). This enabled fast separation and determination of the peptide and all other reactants in one separation with no sample preparation. Covalent RGDS peptide grafting onto the chitosan film surface was demonstrated using solid-state NMR of swollen films. CE indicated that oligomers of RGDS, not simply RGDS, were grafted on the film, with a likely hyperbranched structure. To assess the functional properties of the grafted films, cell growth was compared on control and peptide-grafted chitosan films. Light microscopy and polymerase chain reaction (PCR) analysis demonstrated greatly improved cell attachment to RGDS-grafted chitosan films.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Borsäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Phosphorsäure, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Phosphorsäure, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
N-Hydroxysuccinimid, 98%