Merck
Nephroprotection of plantamajoside in rats treated with cadmium.
Environmental toxicology and pharmacology (2014-12-17)
Ha-Young Jung, Dong-Won Seo, Chung-Oui Hong, Ji-Yeon Kim, Sung-Yong Yang, Kwang-Won Lee
ZUSAMMENFASSUNG
Cadmium (Cd), an environmental and industrial pollutant, generates free radicals responsible for oxidative stress. Cd can also lead to various renal toxic damage such as the proximal tubules and glomerulus dysfunction. Plantamajoside (PMS), a major compound of Plantago asiatica (PA), was reported to have the antioxidant effects. In this study, we investigated the protective effects of PMS on Cd-induced renal damage in the NRK-52E cell and rat kidney tissue. Cd exposure increased the ROS generation, lipid peroxidation, serum biochemical values of renal damage, and mRNA and protein expressions of KIM-1 in vitro and in vivo. The significant reduction in glutathione (GSH)/glutathione disulfide (GSSG) ratio and activities of antioxidant enzymes were also observed in the rats treated with Cd. PMS significantly decreased the ROS generation and lipid peroxidation, thus enhancing GSH/GSSG ratio, antioxidant enzyme activities in the cells and rats, and improved histochemical appearances, indicating that PMS has protective activities against Cd-induced renal injury.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
N-Ethylmaleimid, purum p.a., ≥99.0% (HPLC)
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Triethanolamin, ≥99.0% (GC)
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Triethanolamin, reagent grade, 98%
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
N-Ethylmaleimid, crystalline, ≥98% (HPLC)
Sigma-Aldrich
2-Vinylpyridin, 97%
Sigma-Aldrich
N-Ethylmaleimid, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Cadmiumchlorid, 99.99% trace metals basis
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Glutathion, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutathion reduziert, BioXtra, ≥98.0%
Sigma-Aldrich
Cadmiumchlorid
Sigma-Aldrich
Triethanolamin, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
N-Ethylmaleimid, BioXtra, ≥98% (HPLC)
Supelco
Dimethylsulfoxid, analytical standard
Sigma-Aldrich
2,2′-Bichinolin-4,4′-dicarbonsäure Dinatriumsalz Dihydrat, ≥98% (HPLC)
Sigma-Aldrich
Triethanolamin, puriss. p.a., ≥99% (GC)