Merck
  • Bioactivity-guided isolation and structural characterization of the antifungal compound, plumbagin, from Nepenthes gracilis.
Bioactivity-guided isolation and structural characterization of the antifungal compound, plumbagin, from Nepenthes gracilis.
Pharmaceutical biology (2014-07-16)
Pei Shing Gwee, Kong Soo Khoo, Hean Chooi Ong, Nam Weng Sit
ZUSAMMENFASSUNG
Despite several phytochemical studies of Nepenthes gracilis Korth (Nepenthaceae), the biological activities of this pitcher plant remain to be explored. This study evaluates the antifungal activity of N. gracilis extracts, isolates, and characterizes its bioactive compound and evaluates the cytotoxicity of the isolated compound. Fresh leaves of N. gracilis were sequentially extracted. The fungistatic and fungicidal activities of the extracts were evaluated against six species of fungi of medical importance using a colorimetric broth microdilution method. The most active extract was fractionated by liquid-liquid partitioning and further purified by a preparative thin layer chromatography. Structural elucidation was carried out using FT-IR, GC-MS, and NMR. Cytotoxicity testing against rhesus monkey kidney epithelial cells (LLC-MK2) was assessed by a neutral red uptake (NRU) assay. The hexane extract, which showed the lowest minimum inhibitory concentration (MIC) and minimum fungicidal concentration (MFC), both at 20 μg/mL against Candida albicans, Issatchenkia orientalis, and Trichophyton mentagrophytes, was subjected to bioactivity-guided fractionation. The isolated compound exhibited potent activity with the MIC values ranging from 2 to 31 μg/mL against all the fungi. The active compound was identified as plumbagin (5-hydroxy-2-methyl-naphthalene-1,4-dione). The 50% cytotoxicity concentration (CC50) of plumbagin was 0.60 μg/mL. The selectivity indices of plumbagin against all the fungi were less than 1.0, indicating that plumbagin is more toxic to mammalian than fungal cells. This study provides information on the antifungal properties of N. gracilis leaf extracts, as well as the antifungal and cytotoxicity properties of plumbagin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Hexan, ReagentPlus®, ≥99%
Sigma-Aldrich
Hexan, suitable for HPLC, ≥95%
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Hexan, HPLC Plus, for HPLC, GC, and residue analysis, ≥95%
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
Hexan, Laboratory Reagent, ≥95%
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, flakes