Merck
  • Composite scaffolds of dicalcium phosphate anhydrate /multi-(amino acid) copolymer: in vitro degradability and osteoblast biocompatibility.
Composite scaffolds of dicalcium phosphate anhydrate /multi-(amino acid) copolymer: in vitro degradability and osteoblast biocompatibility.
Journal of biomaterials science. Polymer edition (2015-01-03)
Qianqian Yao, Jun Ye, Qian Xu, Anchun Mo, Ping Gong
ZUSAMMENFASSUNG
This study aims to evaluate in vitro degradability and osteoblast biocompatibility of dicalcium phosphate anhydrate/multi-(amino acid) (DCPA/MAA) composites prepared by in situ polymerization method. The results revealed that the composites could be slowly degraded in PBS solution, with weight loss of 9.5 ± 0.2 wt.% compared with 12.2 ± 0.2 wt.% of MAA copolymer after eight weeks, and the changes of pH value were in the range of 7.18-7.4 and stabilized at 7.24. In addition, the compressive strength of the composite decreased from 98 to 62 MPa while that of MAA copolymer from 117 to 86 MPa. Furthermore, with non-toxicity demonstrated by 3-[4,5-dimethylthiazol-2-yl]-2,5-diphenyl-tetrazolium bromide assay, the addition of DCPA to the MAA copolymer evidenced an enhancement of osteoblast differentiation and attachment compared with pure MAA materials regarding to alkaline phosphatase activity as well as initial cell adhesion. The results indicated that the DCPA/MAA scaffolds with good osteoblast biocompatibility, degradability, and sufficient strength had promising potential application in bone tissue engineering.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, flakes
Supelco
Natronlauge, 49-51% in water, eluent for IC
Supelco
Natronlauge-Konzentrat, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Acetessigsäure-methylester, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis