Merck
Protective Effect of Liriodendrin Isolated from Kalopanax pictus against Gastric Injury.
Biomolecules & therapeutics (2015-01-17)
Yoon Ah Sohn, Seon A Hwang, Sun Yi Lee, In Young Hwang, Sun Whoe Kim, So Yeon Kim, Aree Moon, Yong Soo Lee, Young Ho Kim, Keum Jee Kang, Choon Sik Jeong
ZUSAMMENFASSUNG
In this study, we investigated the inhibitory activities on gastritis and gastric ulcer using liriodendrin which is a constituent isolated from Kalopanax pictus. To elucidate its abilities to prevent gastric injury, we measured the quantity of prostaglandin E2 (PGE2) as the protective factor, and we assessed inhibition of activities related to excessive gastric acid be notorious for aggressive factor and inhibition of Helicobacter pylori (H. pylori) colonization known as a cause of chronic gastritis, gastric ulcer, and gastric cancer. Liriodendrin exhibited higher PGE2 level than rebamipide used as a positive control group at the dose of 500 μM. It was also exhibited acid-neutralizing capacity (10.3%) and H(+)/K(+)-ATPase inhibition of 42.6% (500 μM). In pylorus-ligated rats, liriodendrin showed lower volume of gastric juice (4.38 ± 2.14 ml), slightly higher pH (1.53 ± 0.41), and smaller total acid output (0.47 ± 0.3 mEq/4 hrs) than the control group. Furthermore liriodendrin inhibited colonization of H. pylori effectively. In vivo test, liriodendrin significantly inhibited both of HCl/EtOH-induced gastritis (46.9 %) and indomethacin-induced gastric ulcer (46.1%). From these results, we suggest that liriodendrin could be utilized for the treatment and/or protection of gastritis and gastric ulcer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Trypanblau, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Trypanblau -Lösung, 0.4%, suitable for microscopy
Sigma-Aldrich
Trypanblau, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Stickstoff, ≥99.998%
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Supelco
Ampicillin
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Cimetidin
USP
Ampicillin, United States Pharmacopeia (USP) Reference Standard
Supelco
Cimetidin, Pharmaceutical Secondary Standard; Certified Reference Material
Ampicillin, anhydrous, European Pharmacopoeia (EP) Reference Standard
Dinoproston, European Pharmacopoeia (EP) Reference Standard
USP
Cimetidin, United States Pharmacopeia (USP) Reference Standard
Cimetidin, European Pharmacopoeia (EP) Reference Standard
Cimetidin für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Cimetidin für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard