Merck
  • Influence of Fungal Strain, Temperature, and Wetness Duration on Infection of Grapevine Inflorescences and Young Berry Clusters by Botrytis cinerea.
Influence of Fungal Strain, Temperature, and Wetness Duration on Infection of Grapevine Inflorescences and Young Berry Clusters by Botrytis cinerea.
Phytopathology (2014-10-30)
Nicola Ciliberti, Marc Fermaud, Luca Languasco, Vittorio Rossi
ZUSAMMENFASSUNG
The effect of temperature and wetness duration on infection of Vitis vinifera inflorescences (from "inflorescence clearly visible" to "end of flowering" stages) and young berry clusters (at "fruit swelling" and "berries groat-sized" stages) by Botrytis cinerea was investigated. Artificial inoculations were carried out using conidial suspensions of eight B. cinerea strains belonging to the transposon genotypes transposa and vacuma. Infection incidence was significantly affected by strain but not by transposon genotype (transposon genotype accounted for only 6.5% of the variance). Infection incidence was also affected by the interaction between strain and growth stage of the inflorescence or berry cluster (overall accounting for approximately 57% of the experimental variance). Thus, under our experimental conditions, the ability to cause infection was a strain rather than a transposon genotype attribute. Across all strains, infection incidence was lowest when inflorescences were clearly visible or fully developed, highest at flowering (from beginning to end of flowering), and intermediate at the postflowering fruit stages (fruit swelling and berries groat-sized). One transposa strain, however, was highly virulent on all grapevine growth stages tested. The effects of temperature and wetness duration on infection incidence were similar for all fungal strains and grapevine growth stages; infection incidence was highest at 20°C and lowest at 30°C, and was also low at 5°C. Similar results were obtained for mycelial growth and conidial germination. Based on the pooled data for all strains and grapevine growth stages, an equation was developed that accounted for the combined effects of temperature and wetness duration on relative infection incidence. This equation should be useful for developing decision-making systems concerning B. cinerea control at early grapevine growth stages.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Phenol -Lösung, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Milchsäure, meets USP testing specifications
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
Calciumhypochlorit, technical grade
Sigma-Aldrich
Milchsäure, 88%, FCC
Sigma-Aldrich
L-Lysin -monohydrochlorid, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol -Lösung, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Milchsäure, natural, ≥85%