Merck
Blood components and OCT reflectivity evaluated in animal model.
Current eye research (2014-10-14)
Shozo Sonoda, Taiji Sakamoto, Makoto Shirasawa, Takehiro Yamashita, Eisuke Uchino, Hiroto Terasaki
ZUSAMMENFASSUNG
To see the relationship between blood components and optical coherence tomography (OCT) reflectivity using an animal model in which the aqueous humor was substituted by different experimental solutions without changing the integrity of the retina. The aqueous humor of an enucleated swine eye was replaced with plasma obtained from healthy volunteers. The OCT reflectivity of the anterior chamber filled with each plasma was calculated from individual OCT images, and was expressed by an arbitrary unit (AU). The concentration of blood components such as cholesterol, hemoglobin (Hb) and bilirubin of each individual was measured, and the correlation between each of them and the OCT reflectivity of aqueous humor in an enucleated swine eye was analyzed. Using the same model, the effects of the single plasma component on OCT reflectivity were examined. Blood samples were obtained from 24 individuals. OCT reflectivity was 30.68 ± 14.8 AU (average ± SD), ranging from 11.11 to 60.31 AU. OCT reflectivity correlated significantly with the concentration of triglycerides (R = 0.634, p = 0.001) and total cholesterol (R = 0.488, p = 0.015) using Spearman's rank correlation coefficient. While a partial correlation analysis showed that it correlated significantly with triglyceride (R = 0.60, p = 0.003), but not total cholesterol. OCT reflectivity was highest in a balanced salt solution (BSS) with Hb (average 42.05 AU), followed by fibrinogen (8.08 AU), bilirubin (6.12 AU) and γ-globulin (2.85 AU). Albumin did not increase the reflectivity of the BSS with a normal concentration (1.11 AU) compared to the control BSS alone (0.73 AU). OCT reflectivity was most strongly affected by the presence of triglycerides among the blood components. Some molecules such as Hb and fibrinogen significantly increase the OCT reflectivity. This information should be helpful for interpreting the OCT findings correctly.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.