Merck
  • On the use of aptamer microarrays as a platform for the exploration of human prothrombin/thrombin conversion.
On the use of aptamer microarrays as a platform for the exploration of human prothrombin/thrombin conversion.
Analytical biochemistry (2015-01-15)
Camille Daniel, Yoann Roupioz, Thierry Livache, Arnaud Buhot
ZUSAMMENFASSUNG
Microarrays are particular biosensors with multiple grafted probes that are generally used for parallel and simultaneous detection of various targets. In this study, we used microarrays with aptamer probes in order to follow up the different biomolecular interactions of a single enzyme, the thrombin protein, involved in the complex coagulation cascade. More precisely, thanks to label-free surface plasmon resonance imaging, we were able to monitor in real time an important step in the firing of the coagulation cascade in situ-the enzymatic transformation of prothrombin into thrombin, catalyzed by factor Xa. We were also able to appraise the influence of other biochemical factors and their corresponding inhibiting or enhancing behaviors on thrombin activation. Our study opens the door for the development of a complete microarray-based platform not only for the whole coagulation cascade analysis but also for novel drug screening assays in pharmacology.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethylformamid, ACS reagent, ≥99.8%
Sigma-Aldrich
N,N-Dimethylformamid, suitable for HPLC, ≥99.9%
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
N-Hydroxysuccinimid, 98%
Sigma-Aldrich
N,N-Dimethylformamid, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Sigma-Aldrich
N,N-Dimethylformamid, biotech. grade, ≥99.9%
Sigma-Aldrich
11-Mercaptoundecansäure, 95%
Supelco
Dimethylformamid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
N,N-Dimethylformamid, analytical standard
Sigma-Aldrich
11-Mercaptoundecansäure, 98%
Sigma-Aldrich
N-Hydroxysuccinimid, purum, ≥97.0% (T)
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar