Merck
  • Hypercholesterolemia increases plasma saturated and n-6 fatty acids altering prostaglandin homeostasis and promotes endothelial dysfunction in rabbits.
Hypercholesterolemia increases plasma saturated and n-6 fatty acids altering prostaglandin homeostasis and promotes endothelial dysfunction in rabbits.
Lipids (2014-06-02)
M Medina, M R Alberto, L Sierra, C Van Nieuwenhove, S Saad, M I Isla, S Jerez
ZUSAMMENFASSUNG
The present study evaluated the plasma fatty acid levels and the vascular prostaglandin (PG) release in a rabbit model of early hypercholesterolemia with endothelial dysfunction. Rabbits were fed either a control diet (CD) or a diet containing 1 % cholesterol (HD) for 5-6 weeks. The level of fatty acids was measured in plasma. The levels of PG and nitric oxide (NO) released from the aorta were also determined. Vascular morphology of the aorta was characterized by intima and media thickness measurements. The rabbits fed with HD had higher levels of arachidonic acid (ARA) and lower levels of oleic acid. The linoleic acid level was unchanged. PGI(2) and NO were diminished and PGF(2α) levels, the PGI(2)/TXA(2) ratio and the intima/media ratio were increased in rabbits fed with HD. In conclusion, feeding HD for a short period increased ARA plasma levels and unbalanced release of vasodilator/vasoconstrictor PG redirected the pathway to vasoconstrictor metabolite release. These lipid metabolism alterations in addition to the reduced NO levels and the moderate changes in the vascular morphology contributed to the endothelial dysfunction in this animal model. Therefore, the present findings support the importance of early correction or prevention of high cholesterol levels to disrupt the endothelial dysfunction process that leads to cardiovascular disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.