Merck
Alpha-amylase reactivity in relation to psychopathic traits in adults.
Psychoneuroendocrinology (2015-02-11)
Andrea L Glenn, Rheanna J Remmel, Adrian Raine, Robert A Schug, Yu Gao, Douglas A Granger
ZUSAMMENFASSUNG
Recent investigations of the psychobiology of stress in antisocial youth have benefited from a multi-system measurement model. The inclusion of salivary alpha-amylase (sAA), a surrogate marker of autonomic/sympathetic nervous system (ANS) activity, in addition to salivary cortisol, a biomarker of the hypothalamic-pituitary-adrenal (HPA) axis functioning, has helped define a more complete picture of individual differences and potential dysfunction in the stress response system of these individuals. To the authors' knowledge, no studies have examined sAA in relation to antisocial behavior in adults or in relation to psychopathic traits specifically. In the present study, we examined sAA, in addition to salivary cortisol, in a relatively large sample (n=158) of adult males (M age=36.81, range=22-67 years; 44% African-American, 34% Caucasian, 16% Hispanic) recruited from temporary employment agencies with varying levels of psychopathic traits. Males scoring highest in psychopathy were found to have attenuated sAA reactivity to social stress compared to those scoring lower in psychopathy. No differential relationships with the different factors of psychopathy were observed. In contrast to studies of antisocial youth, there were no interactions between sAA and cortisol levels in relation to psychopathy, but there was a significant interaction between pre-stressor levels of sAA and cortisol. Findings reveal potential regulatory deficits in the fast-acting, 'fight or flight', component of the stress response in adult males with psychopathic traits, as well as abnormalities in how this system may interact with the HPA axis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Hydrocortison, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortison, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Maltotriose, ≥90% (HPLC)
Sigma-Aldrich
Hydrocortison, ≥98% (HPLC)
Supelco
Hydrocortison, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Hydrocortison, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrocortison, meets USP testing specifications
Sigma-Aldrich
2-Chlor-4-Nitrophenol, 97%
Hydrocortison für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Hydrocortison, European Pharmacopoeia (EP) Reference Standard
Hydrocortison, British Pharmacopoeia (BP) Assay Standard