Merck
Cellular localization and detergent dependent oligomerization of rice allene oxide synthase-1.
Journal of plant research (2014-10-20)
Sereyvath Yoeun, Jeong-Il Kim, Oksoo Han
ZUSAMMENFASSUNG
Allene oxide synthase-1 from Oryza sativa (OsAOS1) localizes to the chloroplast, but lacks a putative chloroplast targeting sequence typically found in dicot AOS. Here, kinetic parameters and the oligomerization state/subunit composition of OsAOS1 were characterized in vitro in the absence or presence of detergent micelles. The catalytic efficiency (k(cat)/K(m)) of OsAOS1 reached a maximum near the critical micelle concentration for polyoxyethylene 10 tridecyl ether. Native gel analysis showed that OsAOS1 exists as a multimer in the absence of detergent micelles. The multimeric form of OsAOS1 was stably cross-linked in the absence of detergents, while only monomeric OsAOS1 was detected in the presence of detergent micelles. Gel filtration analysis indicated that the oligomeric state of OsAOS1 depends strongly on the detergents and that the monomer becomes the predominant form in the presence of detergent micelles. These data suggest that the detergent-dependent oligomeric state of OsAOS1 is an important factor for the regulation of its catalytic efficiency.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,2-Dichlorethan, ACS reagent, ≥99.0%
Sigma-Aldrich
1,2-Dichlorethan, anhydrous, 99.8%
Sigma-Aldrich
Vitamin B12, ≥98%
Supelco
Cyanocobalamin, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Supelco
Cyanocobalamin (B12), analytical standard
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Supelco
1,2-Dichlorethan, analytical standard
Cyanocobalamin, British Pharmacopoeia (BP) Assay Standard
Cyanocobalamin, European Pharmacopoeia (EP) Reference Standard