Merck
Are human adenovirus-5 and 36 associated with obesity in children?
Journal of investigative medicine : the official publication of the American Federation for Clinical Research (2014-07-06)
Elcin Kal Cakmakliogullari, Tamer Sanlidag, Betul Ersoy, Sinem Akcali, Ahmet Var, Candan Cicek
ZUSAMMENFASSUNG
The aims of this study were to determine the association between adenovirus-5- and adenovirus-36-specific antibodies and obesity in children and to investigate their relationship with serum lipid and leptin levels. A cross-sectional study was performed on a total of 120 children who were divided into subgroups according to body mass index percentile as obese (≥ 95th percentile) or nonobese (<95th percentile). The presence of adenovirus-36 and adenovirus-5-neutralizing antibodies was investigated by using the serum neutralization assay. Serum leptin levels were determined by microenzyme immonoassay; high-density lipoprotein, low-density lipoprotein, triglyceride, and cholesterol levels were measured by chemiluminescence method. The presence of adenovirus-5-specific antibodies was 28.3% and 6.6% in the obese children and in non-obese children, respectively (P = 0.02). The frequency of adenovirus-36-specific antibodies was significantly greater (P = 0.018) in the obese children (26.6%) than in the non-obese children (10.0%). Serum leptin level of the obese group were significantly higher than that of the non-obese group (P = 0.000). Our data support the association between obesity and the presence of specific antibodies to adenovirus-36 and adenovirus-5 in children. Our research has the feature of being the first national study to indicate the relationship between adenovirus-36 and human obesity as well as the first international study to indicate the relationship between adenovirus-5 and human obesity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Supelco
Cholesterin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterin -Lösung, certified reference material, 10 mg/mL in chloroform
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterin, tested according to Ph. Eur.