Merck
  • Preparation of monolithic molecularly imprinted polymer sol-gel packed tips for high-throughput bioanalysis: extraction and quantification of L-tyrosine in human plasma and urine samples utilizing liquid chromatography and tandem mass spectrometry.
Preparation of monolithic molecularly imprinted polymer sol-gel packed tips for high-throughput bioanalysis: extraction and quantification of L-tyrosine in human plasma and urine samples utilizing liquid chromatography and tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-08-12)
Mohammad Mahdi Moein, Aziza El-Beqqali, Abbi Abdel-Rehim, Amin Jeppsson-Dadoun, Mohamed Abdel-Rehim
ZUSAMMENFASSUNG
In situ monolithic molecularly imprinted polymer sol-gel packed tips (MMSTs) were prepared and evaluated for the extraction of lung cancer biomarker L-tyrosine (Tyr) from human plasma and urine samples. Several extraction parameters such as the conditioning, washing and elution solutions, pH and time were investigated. The enrichment factor (EF) and extraction recovery (ER) were studied. MMST showed good selectivity and a high extraction recovery, and MMST as a sorbent showed good stability and repeatability. The method validation showed good regression correlation coefficients for plasma and urine samples (R(2)≥0.996) within the concentration range of 5-1000 and 1-1000 nmol L(-1) in plasma and urine samples, respectively. The lower limits of quantification (LLOQ) in the plasma and urine samples were 5 and 1 nmol L(-1), respectively. The between-batch precision for Tyr in plasma ranged from 1.0 to 6.0%, and in urine it was from 1.0 to 7.0%. The results show that the developed method has more facility, stability, durability and repeatability compared with previous similar methods. To the best of our knowledge, this is the first study aimed at the selective separation of Tyr as a lung cancer biomarker by MMSTs from biological matrixes and detection by LC/MS/MS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoressigsäure, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
3-(Trimethoxysilyl)propylmethacrylat, 98%
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
L-Tyrosin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
3-(Trimethoxysilyl)propylmethacrylat, ≥97%
SAFC
Glycin
Supelco
Trifluoressigsäure, analytical standard
Sigma-Aldrich
Sarcosin, 98%
Supelco
Glycin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Tyrosin, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
L-Tyrosin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Hippursäure, 98%
Sigma-Aldrich
Glycin, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycin, ACS reagent, ≥98.5%
SAFC
L-Tyrosin
Sigma-Aldrich
Glycin, 99%, FCC
Supelco
L-Tyrosin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycin, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
USP
Glycin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycin, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
USP
L-Tyrosin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
L-Tyrosin, FG
Sigma-Aldrich
Sarcosin, BioXtra
Sigma-Aldrich
Sarcosin, crystallized, ≥98.0% (T)