Merck
  • COX-2 mediated induction of endothelium-independent contraction to bradykinin in endotoxin-treated porcine coronary artery.
COX-2 mediated induction of endothelium-independent contraction to bradykinin in endotoxin-treated porcine coronary artery.
Journal of cardiovascular pharmacology (2014-09-06)
Amar S More, Hye Min Kim, Ru Zhao, Gilson Khang, Tobias Hildebrandt, Christian Bernlöhr, Henri Doods, Dongwon Lee, Seung Hee Lee, Paul M Vanhoutte, Dongmei Wu
ZUSAMMENFASSUNG
This study examined the vascular effects of bradykinin in health and vascular inflammation comparing responses of isolated pig coronary arteries in the absence and presence of endotoxins. Bradykinin induced contractions in lipopolysaccharide-treated, but not untreated, arterial rings without endothelium. The B2-receptor antagonist HOE140, but not the B1-receptor inhibitor SSR240612, blocked these endothelium-independent contractions in response to bradykinin. The bradykinin-induced contractions were blocked by indomethacin, celecoxib, and terbogrel but not valeryl salicylate, AH6809, AL 8810, or RO1138452. They were attenuated by N-(p-amylcinnamoyl) anthranilic acid, and by diethyldithiocarbamate plus tiron but not by L-NAME. Quantitative reverse-transcription polymerase chain reaction revealed significant upregulations of messenger RNA expressions of B1 receptors, COX-2, and thromboxane A synthase 1 (TBXAS1) following lipopolysaccharide incubation but not of B2 receptors or COX-1. The present data demonstrate that bradykinin induces contractions mediated by the COX-2 pathway in endotoxin-treated pig coronary arteries. These results support differential roles of bradykinin in health and disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Indomethacin, 98.0-102.0%, meets EP testing specifications
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Anthranilsäure, reagent grade, ≥98%
Sigma-Aldrich
Stickstoff, ≥99.998%
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Indomethacin, meets USP testing specifications
Sigma-Aldrich
Anthranilsäure, puriss. p.a., ≥99.5% (T)
Supelco
Indomethacin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
9,11-Dideoxy-11α,9α-Epoxymethanoprostaglandin F -Lösung, solution, 10 mg/mL in methyl acetate
Natriumlaurylsulfat, European Pharmacopoeia (EP) Reference Standard
USP
Indomethacin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Celecoxib, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Indomethacin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Valeryl Salicylate, ≥98% (HPLC)