Merck
Antinociceptive effect of ethanol extract of leaves of Lannea coromandelica.
Journal of ethnopharmacology (2014-03-26)
Mohammad Zafar Imam, Md Moniruzzaman
ZUSAMMENFASSUNG
Lannea coromandelica (Houtt.) Merr. is a plant locally called "Jiga", found all over Bangladesh. Leaf of the plant is traditionally used in the treatment of local swellings, pains of body, toothache etc. This study evaluated the antinociceptive effect of the ethanol extract of L. coromandelica leaves (EELC). The antinociceptive activity of the extract (at the doses of 50, 100, and 200 mg/kg) was evaluated by using chemical- and heat-induced pain models such as acetic acid-induced writhing, hot plate, tail immersion, formalin, and glutamate test. To verify the possible involvement of opioid receptor in the central antinociceptive effect of EELC, naloxone was used to antagonize the effect. Besides, the involvements of ATP-sensitive K(+) channel and cGMP pathway were also justified by using glibenclemide and methylene blue. EELC demonstrated significant dose-dependent antinociceptive activity in the chemical- and heat-induced nociception in mice models (p<0.05). These findings imply the involvement of both peripheral and central antinociceptive mechanisms. The use of naloxone confirmed the association of opioid receptors in the central antinociceptive effect. EELC also showed the involvements of ATP-sensitive K(+) channel and cGMP pathway for antinociceptive activity. This study reported the antinociceptive activity of the leaf of L. coromandelica and rationalized the traditional use of the leaf in the treatment of different painful conditions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Essigsäure, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Essigsäure, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyd -Lösung, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Essigsäure, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Essigsäure -Lösung, suitable for HPLC
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
L-Glutaminsäure, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Ethanol, BioUltra, Molecular Biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Methylenblau, certified by the BSC
Sigma-Aldrich
L-Glutaminsäure, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Methylenblau -Lösung, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Supelco
Ethanol, wasserfrei, Pharmaceutical Secondary Standard; Certified Reference Material