Merck
Prevention of obesity-induced renal injury in male mice by DPP4 inhibition.
Endocrinology (2014-04-10)
Ravi Nistala, Javad Habibi, Guido Lastra, Camila Manrique, Annayya R Aroor, Melvin R Hayden, Mona Garro, Alex Meuth, Megan Johnson, Adam Whaley-Connell, James R Sowers
ZUSAMMENFASSUNG
Therapies to prevent renal injury in obese hypertensive individuals are being actively sought due to the obesity epidemic arising from the Western diet (WD), which is high in fructose and fat. Recently, activation of the immune system and hyperuricemia, observed with high fructose intake, have been linked to the pathophysiology of hypertension and renal injury. Because dipeptidyl peptidase 4 (DPP4) is a driver of maladaptive T-cell/macrophage responses, renal-protective benefits of DPP4 inhibition in the WD-fed mice were examined. Mice fed a WD for 16 weeks were given the DPP4 inhibitor MK0626 in their diet beginning at 4 weeks of age. WD-fed mice were obese, hypertensive, and insulin-resistant and manifested proteinuria and increased plasma DPP4 activity and uric acid levels. WD-fed mice also had elevated kidney DPP4 activity and monocyte chemoattractant protein-1 and IL-12 levels and suppressed IL-10 levels in the kidney, suggesting macrophage-driven inflammation, glomerular and tubulointerstitial injury. WD-induced increases in DPP4 activation in the plasma and kidney and proteinuria in WD mice were abrogated by MK0626, although blood pressure and systemic insulin sensitivity were not improved. Contemporaneously, MK0626 reduced serum uric acid levels, renal oxidative stress, and IL-12 levels and increased IL-10 levels, suggesting that suppression of DPP4 activity leads to suppression of renal immune/inflammatory injury responses to a WD. Taken together, these results demonstrate that DPP4 inhibition prevents high-fructose/high-fat diet-induced glomerular and tubular injury independent of blood pressure/insulin sensitivity and offers a potentially novel therapy for diabetic and obesity-related kidney disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Supelco
Natriumchlorid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Natriumchlorid, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, tested according to Ph. Eur.
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis