Merck
Peroxynitrite and protein nitration in the pathogenesis of diabetic peripheral neuropathy.
Diabetes/metabolism research and reviews (2014-04-02)
Roman Stavniichuk, Hanna Shevalye, Sergey Lupachyk, Alexander Obrosov, John T Groves, Irina G Obrosova, Mark A Yorek
ZUSAMMENFASSUNG
Peroxynitrite, a product of the reaction of superoxide with nitric oxide, causes oxidative stress with concomitant inactivation of enzymes, poly(ADP-ribosylation), mitochondrial dysfunction and impaired stress signalling, as well as protein nitration. In this study, we sought to determine the effect of preventing protein nitration or increasing peroxynitrite decomposition on diabetic neuropathy in mice after an extended period of untreated diabetes. C57Bl6/J male control and diabetic mice were treated with the peroxynitrite decomposition catalyst Fe(III) tetramesitylporphyrin octasulfonate (FeTMPS, 10 mg/kg/day) or protein nitration inhibitor (-)-epicatechin gallate (20 mg/kg/day) for 4 weeks, after an initial 28 weeks of hyperglycaemia. Untreated diabetic mice developed motor and sensory nerve conduction velocity deficits, thermal and mechanical hypoalgesia, tactile allodynia and loss of intraepidermal nerve fibres. Both FeTMPS and epicatechin gallate partially corrected sensory nerve conduction slowing and small sensory nerve fibre dysfunction without alleviation of hyperglycaemia. Correction of motor nerve conduction deficit and increase in intraepidermal nerve fibre density were found with FeTMPS treatment only. Peroxynitrite injury and protein nitration are implicated in the development of diabetic peripheral neuropathy. The findings indicate that both structural and functional changes of chronic diabetic peripheral neuropathy can be reversed and provide rationale for the development of a new generation of antioxidants and peroxynitrite decomposition catalysts for treatment of diabetic peripheral neuropathy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Glycin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycin, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Glycin, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%