Merck
  • Preparation of Keggin-type phosphomolybdate by a one-step solid-state reaction at room temperature and its application in protein adsorption.
Preparation of Keggin-type phosphomolybdate by a one-step solid-state reaction at room temperature and its application in protein adsorption.
Journal of separation science (2014-07-22)
Qing Chen, Li-Ming Shen, Jie Xia, Xu-Wei Chen, Jian-Hua Wang
ZUSAMMENFASSUNG
Keggin-type phosphomolybdate ((C19H42N)3PMo12O40) is prepared by a one-step solid-state reaction at room temperature and characterized by scanning electron microscopy, Fourier transform infrared spectroscopy, X-ray diffraction, thermogravimetric analysis, and elemental analysis. The as-prepared phosphomolybdate is demonstrated to be an efficient adsorbent for proteins. In this particular case, the selective adsorption of neutral protein hemoglobin is achieved. While under the same conditions virtually no adsorption of acidic and basic proteins, represented by bovine serum albumin and cytochrome c, are observed. A solid-phase extraction procedure is developed for the selective isolation of hemoglobin. At pH 6, a sorption efficiency of 91.4% is achieved for 100 μg/mL hemoglobin in 1.0 mL solution by using 5.0 mg of the phosphomolybdate. The adsorption behavior of hemoglobin fits well with a Langmuir adsorption model, corresponding to a theoretical adsorption capacity of 55.86 mg/g. The retained hemoglobin could be readily recovered by using a 60 mmol/L imidazole solution at pH 7, giving rise to a recovery of 64.7%. The practical application of phosphomolybdate for protein adsorption is demonstrated by the selective isolation of hemoglobin from human whole blood followed by a sodium dodecyl sulfate polyacrylamide gel electrophoresis assay.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Natriumbicarbonat, ACS reagent, ≥99.7%
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Natriumcarbonat, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Natriumcarbonat, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Natriumbicarbonat, ReagentPlus®, ≥99.5%, powder
Sigma-Aldrich
Imidazol, ReagentPlus®, 99%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Zitronensäure, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Imidazol, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumcarbonat, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Zitronensäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumcarbonat, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Imidazol, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Zitronensäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Zitronensäure, 99%
Sigma-Aldrich
Natriumbicarbonat, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%