Merck
  • Type 2 diabetes mellitus control and atherosclerosis prevention in a non-obese rat model using duodenal-jejunal bypass.
Type 2 diabetes mellitus control and atherosclerosis prevention in a non-obese rat model using duodenal-jejunal bypass.
Experimental and therapeutic medicine (2014-08-15)
Xuan Chen, Zhen Huang, Wenhua Ran, Gang Liao, Lang Zha, Ziwei Wang
ZUSAMMENFASSUNG
Type 2 diabetes mellitus (T2DM) is a prevalent disease worldwide and during its conventional treatment, vascular complications remain unavoidable. Roux-en-Y gastric bypass (GBP) is able to induce the remission of T2DM. However, studies of duodenal-jejunal bypass (DJB), a modified procedure of GBP, are being carried out to investigate its ability to induce the remission of T2DM and protect the aorta from atherosclerosis. The present study aimed to investigate the effect of DJB on the rate of T2DM remission and the prevention of atherosclerosis in the aorta in rats with streptozotocin-induced diabetes without obesity, and to explore the mechanism of DJB in protecting the aorta from atherosclerosis. A T2DM rat model was established with a high-fat diet and low-dose streptozotocin. Surgery was performed to analyze its effects on glucose homeostasis, lipid metabolism, inflammation and pathological changes. Furthermore, changes in c-jun NH
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Streptozocin, ≥75% α-anomer basis, ≥98% (HPLC), powder