Merck
  • Microbeads: a novel multiparticulate drug delivery technology for increasing the solubility and dissolution of celecoxib.
Microbeads: a novel multiparticulate drug delivery technology for increasing the solubility and dissolution of celecoxib.
Pharmaceutical development and technology (2013-11-29)
Bernard F McDonald, Ivan S Coulter, Ian W Marison
ZUSAMMENFASSUNG
The purpose of this study was to develop a novel multipaticulate drug delivery technology suitable for the delivery of pre-solubilized celecoxib to the gastrointestinal tract and more specifically to the colon. The solubility of celecoxib in a range of oils, surfactants and co-solvents was evaluated. Celecoxib was solubilized in mixtures of these vehicles to produce liquid formulations. The in vitro dissolution of these liquid formulations was assessed and the data obtained was used to design microbead formulations containing celecoxib dissolved within an emulsion/micellar solution core. Microbead formulations were optimized to increase drug loading, avoid precipitation and to achieve good in vitro dissolution performance. An optimized formulation with a celecoxib loading of 6% w/w was produced and yielded an in vitro dissolution result of 80% over 6 h. The structure of these microbead formulations was characterized using light microscopy to reveal a correlation between droplet size and dissolution performance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Supelco
α-Tocopherylacetat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-α-Tocopherolacetat, ≥96% (HPLC)
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Supelco
Natriumdodecylsulfat, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Supelco
DL-alpha-Tocopherolacetat, analytical standard
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
(+)-α-Tocopherol acetate, oil or semi-solid, ~1360 IU/g
Sigma-Aldrich
(+)-α-Tocopherol acetate, BioReagent, suitable for insect cell culture, ~1360 IU/g
USP
α-Tocopherylacetat, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
DL-α-Tocopherolacetat, tested according to Ph. Eur.
α-Tocopherol-acetat, European Pharmacopoeia (EP) Reference Standard
all-rac-α-Tocopherylacetat für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Natriumlaurylsulfat, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri